Catalog Number | Description | For Use With (Equipment) | Max. Pressure | Material |
---|---|---|---|---|
6036.2100 | Method Transfer Kit for Vanquish Core HPLC Systems | Vanquish Core HPLC System | 1300 bar | Stainless Steel |
6036.2302 | Viper Capillary Kit for Vanquish Core HPLC Systems | Vanquish Core HPLC System | 1300 bar | Stainless Steel |
6036.2303A | Viper Capillary Kit for Vanquish Horizon and Vanquish Flex UHPLC Systems | Vanquish Horizon UHPLC System, Vanquish Flex UHPLC System | 1500 bar | MP35N |
Simplify the installation of Thermo Scientific™ Vanquish™ HPLC and UHPLC systems with Thermo Scientific™ Viper™ Fingertight Fitting Systems, available in pre-assembled capillary kits. These fittings are an ideal fit for the fluidic flow path, providing virtually zero dead volume connections from the pump to the detector. Tool-free setup is fast and easy and the kits ensure robust chromatographic performance to improve daily operations.
Vanquish HPLC and UHPLC systems are shipped with a single system capillary kit as standard.