HyperSep™ Javelin Direct-Connect Online SPE Columns
HyperSep™ Javelin Direct-Connect Online SPE Columns
Actual product may vary
Thermo Scientific™
HyperSep™ Javelin Direct-Connect Online SPE Columns
Clean-up and concentrate target analytes in minutes with these direct-connection SPE guard columns, featuring fingertight design for convenience and efficiency.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSorbentDiameter (Metric) Inner
60310-301HyperSep Retain CX2.1 mm
60310-201HyperSep Retain PEP2.1 mm
60310-202HyperSep Retain PEP3.0 mm
60310-302HyperSep Retain CX3.0 mm
60310-401HyperSep Retain AX2.1 mm
60310-402HyperSep Retain AX3.0 mm
Catalog number 60310-301
Price (USD)/ Each
455.53
-
Add to cart
Sorbent:
HyperSep Retain CX
Diameter (Metric) Inner:
2.1 mm
Price (USD)/ Each
455.53
Add to cart
HyperSep™ Javelin Direct-Connect Online SPE Columns
Catalog number60310-301
Price (USD)/ Each
455.53
-
Add to cart
Clean-up and concentrate target analytes in minutes with Thermo Scientific™ HyperSep™ Online SPE Products. Available in several convenient formats, the columns utilize popular HyperSep SPE chemistries. When used with conventional HPLC systems, columns and appropriate switching valves, they provide:
  • Effective removal of contaminants, such as proteins, from samples
  • Fast, effective clean-up and concentration of target compounds
Thermo Scientific HyperSep Javelin Direct-Connect Online SPE Columns feature a unique fingertight design for convenience and efficiency.
HyperSep Javelin Direct-Connect Online SPE Columns are available with the following HyperSep SPE chemistries: 
  • HyperSep Retain™ PEP — for retention of polar and nonpolar analytes
  • HyperSep Retain-CX — for retention of basic and nonpolar analytes
  • HyperSep Retain-AX — for retention of acidic and nonpolar analytes
  • HyperSep Hypercarb™ — for retention of extremely polar analytes
Specifications
DescriptionHyperSep Retain-CX; 2.1 mm ID
For Use With (Application)Online Solid Phase Extraction
For Use With (Equipment)Conventional HPLC System
Product LineHyperSep
SorbentHyperSep Retain CX
TypeOnline Solid Phase Extraction Column
Diameter (Metric) Inner2.1 mm
Unit SizeEach