Catalog Number | Description |
---|---|
60300-488 | Silica 100 mg/1 mL 96 Removable Well Plate, 1 pk |
60300-485 | Silica 10 mg/1 mL 96 Removable Well Plate, 1 pk |
60300-486 | Silica 25 mg/1 mL 96 Removable Well Plate, 1 pk |
60300-487 | Silica 50 mg/1 mL 96 Removable Well Plate, 1 pk |
60307-234 | Silica 100 mg/2 mL 96 Fixed Well Plate, 1 pk |
60307-233 | Silica 50 mg/2 mL 96 Fixed Well Plate, 1 pk |
60307-231 | Silica 10 mg/2 mL 96 Fixed Well Plate, 1 pk |
Thermo Scientific™ HyperSep™ Silica Plates feature a polar sorbent primarily used to retain analytes in nonpolar matrices, and can effectively separate compounds of very similar structure. Typical applications include:
HyperSep SPE Cartridges and Plates
Thermo Scientific™ HyperSep™ solid phase extraction cartridges offer reproducible, reliable sample preparation in a traditional format. Available in a wide range of chemistries and bed weights, the cartridges are ideal for large samples and conform to industry standard configurations, allowing efficient routine processing by vacuum, by positive pressure or centrifugation.
The Thermo Scientific™ HyperSep-96™ system incorporates traditional solid phase extraction sorbents into 96-well plate format providing fast analysis without sacrificing accuracy or reproducibility. The innovative design of HyperSep-96 well plates provides flexibility for both method development and scale adjustment to 96-well without method revalidation. Well plates are customizable, allowing any arrangement of wells with differing sorbent chemistries for simultaneous processing. HyperSep-96 products are either available in packs of 100 individual wells or in pre-assembled plates.