Catalog Number | Total Volume (Metric) Well | No. per Pack | Volume (Metric) Working | Height (Metric) |
---|---|---|---|---|
60180-P130 | 450 μL | 120 Plates | 10 to 400 μL | 14.4 mm |
60180-P100 | 450 μL | 20 Plates | 10 to 400 μL | 14.4 mm |
60180-P102 | 500 μL | 10 Microplates | 20 to 450 μL | 14.5 mm |
60180-P132 | 500 μL | 120 Microplates | 20 to 450 μL | 14.5 mm |
Thermo Scientific™ SureSTART™ Level 1 WebSeal™ 96-Well Round V-Bottom Microtiter Plates are designed for everyday chromatography analysis.
They provide an affordable choice for use with most HPLC/UHPLC instrument types and are ideal for non-regulated laboratories using water and polar solvents.
These 96-well plates are specification certified to ensure autosampler compatible physical dimensions and conform to ANSI-SLAS 4-2004 standards. Seal these well plates with SureSTART WebSeal 96-Well Plate Sealing Mats, 8 mm Diameter, Round, Level 1.
Performance Level 1 Plates – for Everyday Analysis
Lot Traceable
About Thermo Scientific WebSeal Well Plates & Mats
Thermo Scientific WebSeal plates and mats are available in three performance levels: