SMART Digest™ Bulk Fractionation Kits: Protein G
SMART Digest™ Bulk Fractionation Kits: Protein G
Thermo Scientific™
SMART Digest™ Bulk Fractionation Kits: Protein G
Obtain fast, simple, highly reproducible, and sensitive immunocapture using the high loading capacity of the Thermo Scientific SMART Digest Bulk Fractionation Kits: Protein G.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
60115-103SMART Digest Bulk Fractionation Kit, Protein G, Magnetic, with SOLAμ Collection Plate, Soluble
60115-101SMART Digest Bulk Fractionation Kit, Protein G, Non-Magnetic, Soluble
60115-102SMART Digest Bulk Fractionation Kit, Protein G, Non-Magnetic, with SOLAμ Collection Plate, Soluble
60115-104SMART Digest Bulk Fractionation Kit, Protein G, Magnetic, Soluble
60117-101SMART Digest Bulk Fractionation Kit, Protein G, Magnetic, Standalone
Catalog number 60115-103
Price (USD)/ Each
1,842.36
Add to cart
Description:
SMART Digest Bulk Fractionation Kit, Protein G, Magnetic, with SOLAμ Collection Plate, Soluble
Price (USD)/ Each
1,842.36
Add to cart
SMART Digest™ Bulk Fractionation Kits: Protein G
Catalog number60115-103
Price (USD)/ Each
1,842.36
-
Add to cart
Obtain fast, simple, highly reproducible, and sensitive immunocapture using the high loading capacity of the Thermo Scientific™ SMART Digest™ Bulk Fractionation Kits: Protein G. These kits remove the challenges often associated with protein cleanup by providing a simple and easy capture workflow.
SMART Digest Bulk Fractionation Kits: Protein G contain beads that target purification of immunoglobulins due to the specific and strong affinity for binding to the Fab and Fc portions of the molecule. Choose from non-magnetic beads, or magnetic beads which enable automation. The unwanted interferences are then removed leaving a clean sample containing the target molecule. Once complete, perform digestion using SMART Digest enzymes if required.
Specifications
DescriptionSMART Digest Bulk Fractionation Kit, Protein G, Magnetic, with SOLAμ Collection Plate, Soluble
CaptureProtein G
CleavageC-terminal side of lysine and arginine amino acids
EnzymeTrypsin
FormatSMART Digest Soluble Resin, Bulk Magnetic
For Use With (Application)Immunocapture and Protein Digest Sample Preparation
Product LineSMART Digest
TypeImmunocapture
IncludesBulk Magnetic SMART Digest Soluble Resin, Collection Plate, SOLAμ SPE Plate
Unit SizeEach