SMART Digest™ Bulk Fractionation Kits: Protein A
SMART Digest™ Bulk Fractionation Kits: Protein A
Thermo Scientific™
SMART Digest™ Bulk Fractionation Kits: Protein A
Use this kit to for purification of immunoglobulins prior to protein A digestion.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
60114-102SMART Digest Bulk Protein A Fractionation Kit, Non-Magnetic, with SOLAμ Collection Plate, Soluble
60114-101SMART Digest Bulk Protein A Fractionation Kit, Non-Magnetic, Soluble
60114-103SMART Digest Bulk Protein A Fractionation Kit, Magnetic, with SOLAμ Collection Plate, Soluble
60114-104SMART Digest Bulk Protein A Fractionation Kit, Magnetic, Soluble
60116-101SMART Digest Bulk Protein A Fractionation Kit, Magnetic, Standalone
Catalog number 60114-102
Price (USD)/ Each
1,645.53
Add to cart
Description:
SMART Digest Bulk Protein A Fractionation Kit, Non-Magnetic, with SOLAμ Collection Plate, Soluble
Price (USD)/ Each
1,645.53
Add to cart
SMART Digest™ Bulk Fractionation Kits: Protein A
Catalog number60114-102
Price (USD)/ Each
1,645.53
-
Add to cart
Obtain fast, simple, highly reproducible, and sensitive immunocapture using the high loading capacity of the Thermo Scientific™ SMART Digest™ Bulk Fractionation Kits: Protein A. These kits were designed to remove the challenges often associated with the process of protein clean up by providing a simple and easy capture workflow.
The SMART Digest Bulk fractionation Protein A Kits contain beads (either non-magnetic or magnetic, the magnetic allowing automation) which target purification of immunoglobulins due to the specific and strong affinity for binding to the Fc portion of the molecule. The unwanted interferences are then removed leaving a clean sample containing the target molecule. Once completed subsequent digestion can be facilitated with SMART Digest enzymes if required.
Specifications
DescriptionSMART Digest Bulk Protein A Fractionation Kit, Non-Magnetic, with SOLAμ Collection Plate, Soluble
CaptureProtein A
CleavageC-terminal side of lysine and arginine amino acids
EnzymeTrypsin
FormatSMART Digest Soluble Resin, Bulk Non-magnetic
For Use With (Application)Immunocapture and Protein Digest Sample Preparation
Product LineSMART Digest
TypeImmunocapture
IncludesBulk Non-magnetic SMART Digest Soluble Resin, Collection Plate, SOLAμ SPE Plate
Unit SizeEach