HyperSep™ SLE Plates (pH 9)
HyperSep™ SLE Plates (pH 9)
Actual product may vary
Thermo Scientific™
HyperSep™ SLE Plates (pH 9)
A fast and effective sample preparation technique for removal of phospholipids from biological matricies.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
60109-500-2-9W500 mg/2 mL well, 96 well plate pH9
60109-400-2-9W400 mg/2 mL well, 96 well plate pH9
Catalog number 60109-500-2-9W
Price (USD)/ Each
268.10
-
Add to cart
Description:
500 mg/2 mL well, 96 well plate pH9
Price (USD)/ Each
268.10
Add to cart
HyperSep™ SLE Plates (pH 9)
Catalog number60109-500-2-9W
Price (USD)/ Each
268.10
-
Add to cart
Provide a fast and effective sample preparation technique for removal of phospholipids from biological matrices with Thermo Scientific™ HyperSep™ SLE (Solid supported Liquid/Liquid Extraction).
HyperSep SLE plates (pH 9) deals with sample preparation of biological matrices via a simple, efficient and an effective protcol. The focus being the removal of interfering phospholipids which cause a number of issues for end point analysis.
SLE offers a number of advantages over existing sample preparation techniques such as Liquid/Liquid Extraction (LLE) and Protein Precipitation techniques for dealing with biological matrices. In comparison to LLE, SLE offers greater reproducibility and recoveries, lower solvent requirements and prevents emulsification which is often associated with the LLE technique. It also lends itself to full automation which is not possible via LLE. SLE provides improved cleanliness of sample extract and associated increase in sensitivity compared to protein precipitation techniques which are unable to remove phospholipids.
  • Simple, easy-to-use protocol
  • Removal of phospholipids
  • Greater reproducability and recoveries compared to LLE
Specifications
Description500 mg/2 mL well, 96 well plate pH9
Bed Weight500 mg
For Use With (Application)Removal of Phospholipids From Biological Matrices
Product LineHyperSep
SorbentHyperSep Special Treated Diatomite
TypeSLE Plate
Volume (Metric)2 mL
Unit SizeEach