SMART Digest™ Proteinase K Kits
SMART Digest™ Proteinase K Kits
Thermo Scientific™
SMART Digest™ Proteinase K Kits
Obtain fast, simple, highly reproducible, and sensitive immunocapture and protein digestion in a single well for your biomarker studies.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
60109-109SMART Digest Proteinase K Kit with SOLAμ/collection plate
60109-107SMART Digest Proteinase K Kit, with Collection plate
60109-107-BSMART Digest Proteinase K Kit, Bulk Resin option
60109-107-MBSMART Digest Proteinase K Kit, Magnetic Bulk Resin option
60109-108SMART Digest Proteinase K Kit, with Filter/Collection plate
60109-108-BSMART Digest Proteinase K Kit, Bulk Resin option with Filter/Collection plate
60109-108-MBSMART Digest Proteinase K Kit, Magnetic Bulk Resin option with Filter/Collection plate
60109-109-BSMART Digest Proteinase K Kit Bulk Resin option with SOLAμ/collection plate
60109-109-MBSMART Digest Proteinase K Kit Magnetic Bulk Resin option with SOLAμ/collection plate
Catalog number 60109-109
Price (USD)/ Each
1,230.49
Add to cart
Description:
SMART Digest Proteinase K Kit with SOLAμ/collection plate
Price (USD)/ Each
1,230.49
Add to cart
SMART Digest™ Proteinase K Kits
Catalog number60109-109
Price (USD)/ Each
1,230.49
-
Add to cart
Obtain high quality analytical results from protein digests by using Thermo Scientific™ SMART Digest™ Proteinase K Kits. A significant advance in sample preparation for biopharmaceutical research, the kits provide fast and simple protein digestion with high reproducibility, high sensitivity, and high levels of data quality in a format that's compatible with automation. SMART Digest Kits are a significant improvement over current in-solution protein digestion technologies, which are not reproducible, have poor sensitivity, and employ protracted methodologies not amenable to automation.
Proteinase K Protein Digestion
SMART Digest kits enable you to characterize and quantitate proteins faster, easier, and quicker than ever before. This version of the kit uses proteinase K, which cleaves the peptide bond adjacent to the carboxyl group of aliphatic and aromatic amino acids with blocked alpha amino groups.
Easy and Simple
  • Significantly increased speed of sample processing
  • Simple process
  • Easily automate entire protocol for high-throughput processing
Reliable Results
  • Increased reproducibility over existing protocols
  • Higher sensitivity
  • Higher data confidence
Significantly Reduces Sample Preparation Time
Typically it takes less than 60 minutes (dependent on sample complexity) to achieve full digestion, whereas an in-solution digest can take up to 24 hours.
Cleaner Samples
Reduction in the number of chemicals used and the immobilized proteinase K design helps mitigate against chemically induced post translational modifications (PTMs) and autolysis. The results are easier to interpret.
Couple with SOLAμ SPE Plates for Desalting
When used with the SMART Digest for desalting, Thermo Scientific™ SOLAμ™ SPE solid phase extraction plates provide a fast, easy-to-use, robust, and easily automatable sample preparation solution for analysis of biomolecules.
Specifications
DescriptionSMART Digest Proteinase K Kit with SOLAμ/collection plate
CaptureNone
CleavagePeptide bond adjacent to carboxyl group of aliphatic and aromatic amino acids with blocked alpha amino groups
EnzymeProteinase K
FormatSMART Digest Resin Pre-filled into PCR Tubes
For Use With (Application)Peptide Quantitation, Peptide Mapping
Product LineSMART Digest
TypeProtein Digestion
IncludesSMART Digest Resin Pre-filled into 96 PCR Tubes, Collection Plate, SOLAμ SPE Plate
Unit SizeEach