Catalog Number | Description |
---|---|
60109-1000-6-9 | 1 g/6 mL, Pk. of 30, pH9 |
60109-500-6-9 | 500 mg/6 mL, Pk. of 30, pH9 |
60109-500-3-9 | 500 mg/3 mL, Pk. of 50, pH9 |
60109-200-3-9 | 200 mg/3 mL, Pk. of 50, pH9 |
60109-2000-12-9 | 2 g/12 mL, Pk. of 20, pH9 |
60109-20000-60-9 | 20 g/60 mL, Pk. of 10, pH9 |
60109-4000-25-9 | 4 g/25 mL, Pk. of 15, pH9 |
Provide a fast and effective sample preparation technique for removal of phospholipids from biological matrices with Thermo Scientific™ HyperSep™ SLE (Solid supported Liquid/Liquid Extraction).
HyperSep SLE cartrides (pH9) deals with sample preparation of biological matrices via a simple, efficient and an effective protcol. The focus being the removal of interfering phospholipids which cause a number of issues for end point analysis.