Catalog Number | Description | Sorbent |
---|---|---|
60109-001 | SOLA HRP SPE Cartridge | HRP (Reversed Phase Polymeric) |
60409-001 | SOLA HRP SPE Cartridge 30 mg/3 mL | HRP (Reversed Phase Polymeric) |
60109-002 | SOLA SCX SPE Cartridge | Mixed Mode Strong Cation Exchanger, Polymeric |
60409-002 | SOLA SCX SPE Cartridge 30 mg/3 mL | Mixed Mode Strong Cation Exchanger, Polymeric |
60109-003 | SOLA SAX SPE Cartridge | Mixed Mode Strong Anion Exchanger, Polymeric |
60409-003 | SOLA SAX SPE Cartridge 30 mg/3 mL | Mixed Mode Strong Anion Exchanger, Polymeric |
60109-004 | SOLA WCX SPE Cartridge | Mixed Mode Weak Cation Exchanger, Polymeric |
60409-004 | SOLA WCX SPE Cartridge 30 mg/3 mL | Mixed Mode Weak Cation Exchanger, Polymeric |
60109-005 | SOLA WAX SPE Cartridge | Mixed Mode Weak Anion Exchanger, Polymeric |
60409-005 | SOLA WAX SPE Cartridge 30 mg/3 mL | Mixed Mode Weak Anion Exchanger, Polymeric |
SOLA cartridges provide unparalleled performance characteristics compared to conventional SPE, phospholipid removal and protein precipitation products.
What SOLA Cartridges Mean to Your Analysis