Use the safety tray as the bottom of the glass block vacuum manifold to contain spills.
Have Questions?
Catalog number 60104-260
Price (USD)/ Each
-
Request A Quote
Description:
Manifold Safety Tray, 1 pk
Use the Thermo Scientific™ HyperSep™ Glass Block Vacuum Manifold Safety Tray as the bottom of the manifold and to contain spills. The HyperSep Glass Block Vacuum Manifold enables solid phase extraction sample preparation using 16 or 24 SPE cartridges.
Specifications
DescriptionManifold Safety Tray, 1 pk
For Use With (Equipment)Glass Block Vacuum Manifold Adaptors