Multidrop™ Combi nL Reagent Dispenser, 50nL to 50μL
Multidrop™ Combi nL Reagent Dispenser, 50nL to 50μL
Thermo Scientific™

Multidrop™ Combi nL Reagent Dispenser, 50nL to 50μL

Thermo Scientific™ Multidrop™ Combi nL Rreagent Dispenser provides accurate, consistent and high throughput dispensing for all laboratories across a volume range of 50nL to 50μL.
Have Questions?
Catalog number 5840400
Price (USD)/ Each
36,500.00
Online exclusive
40950.00 
Save 4,450.00 (11%)
-
Add to cart
Description:
Multidrop Combi nL low volume reagent dispenser
Price (USD)/ Each
36,500.00
Online exclusive
40950.00 
Save 4,450.00 (11%)
Add to cart
Multidrop™ Combi nL Reagent Dispenser, 50nL to 50μL
Catalog number5840400
Price (USD)/ Each
36,500.00
Online exclusive
40950.00 
Save 4,450.00 (11%)
-
Add to cart

Experience accurate, consistent, high throughput dispensing for all laboratories with the Thermo Scientific™ Multidrop™ Combi nL Reagent Dispenser. The Combi nL is a nano- to microvolume bulk reagent dispenser with a volume range of 50nL to 50μL. It expands the range of the market leading Multidrop product family, offering easy entry to precise, accurate and reliable low-volume dispensing in pharmaceutical and biotechnology laboratories.

Key Features:
  • Offers low volume dispensing for volumes 50nL -50μL into 96-, 384- and 1536-well plates
  • Results in high quality assay data
  • Increased flexibility—dispensing repeatably into plates with variable height allowing the user to fill either rows, columns, or wells with the easy to use FILLit Software
  • Effortless dispensing—the visual, icon-based onboard user interface makes all functions easy to set tup, use, and maintain
  • Reduced reagent costs—low volume dispensing, a minimal dead volume and the backflush function minimize the use of expensive reagents
  • High throughput—fast dispensing combined with full robotic compatibility ensures increased assay throughput for laboratories needing low volume assay formats

Recommended for:

Assay development; Primary and secondary screening; Genomics and proteomics research; PCR set-up; Sequencing set-up; Cell based assays

Specifications
Accuracy<1μL: ±5%
>1μL: ±2%
Product TypeReagent Dispenser
DescriptionMultidrop Combi nL low volume reagent dispenser
Volume (Metric)50 nL to 50 μL
Increments1 nL (50 to 999 nL), 10 nL (1.00 to 9.99 μL), 100 nL (10.0 to 50.0 μL), 1 nL (FILLit Software)
InterfaceSerial RS-232, USB
Plate Type96-, 384- and -1536-well plates
Precision50nL: CV≤10%, 0.5μL:CV≤5%, 1-10μL:CV≤4%, >10μL: CV≤2%
Speed50 nL in 6 sec., 1 μL in 8 sec. (384-well plate) and 50 nL in 21 sec., 1 μL in 27 sec. (1536-well plate)
Volume (Metric) Dead<1.2 mL
Width (English)14 in.
Width (Metric)35.5 cm
Height (English)8.6 in.
Height (Metric)22 cm
Depth (English)14.8 in.
Depth (Metric)37.5 cm
Weight (English)21.2 lb.
Weight (Metric)9.6 kg
Unit SizeEach