Priced right, the HAAKE Mini CTW Micro-Conical Twin Screw Compounder is the answer: The HAAKE MiniCTW requires as little as five grams or seven milliliters (ml) of material for compounding, which is especially beneficial for industries that compound expensive or small-scale materials such as nano-composites, bio-polymers or pharmaceuticals. By using the optional force feeder, customers can continuously extrude very small material volumes. The instrument is based on proven, conical twin screw technology with co- and counter-rotating screws, and it works as a standalone unit with data export. Due to the channel and a backflow bypass valve, the residence time can be exactly defined.
The HAAKE MiniCTW Micro-Conical Twin Screw Compounder is ideal for the precisely controlled reactive extrusion of high-viscous melts that is being done via an MS-Windows™based software that offers several test set-ups. By running the instrument in circulation mode, the required reaction time for the reactive mixture can be controlled easily. At the end of the test, the bypass valve can be opened and the sample is extruded as a strand. By measuring the torque of the drive motor, the reaction process can be monitored effectively. Test results are stored in the software. The HAAKE MiniCTW Micro-Conical Twin Screw Compounder is one model within our suite of micro-conical twin screw compounders that serves a variety of markets, including polymer, bioscience, nanotechnology and pharmaceutical. The instrument differs from the well-established Thermo Scientific HAAKE MiniLab micro-compounder in that it is designed specifically for industrial research and development whereas the HAAKE MiniLab is focused on polymer development applications. To complete our comprehensive solutions, our international team of material characterization experts provides proven answers for small sample compounding.
Recommended for:
This miniaturized, high-tech tool is perfect for in industrial research in material science in the following markets: