Cryo Vial Closure Color Coders
Cryo Vial Closure Color Coders
Thermo Scientific™
Cryo Vial Closure Color Coders
Make it easier to identify specific vials quickly and simplify your inventory by using these color coders that fit into the top of cryogenic vial or tube closures.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionColorUnit Size
5045-0005Nalgene™ Color coderRedCase of 500
354755Nunc™ CryoColor Vial Closure Coder, PSWhiteCase of 2000
354755PK
also known as 354755
Nunc™ CryoColor Vial Closure Coder, PSWhitePack of 500
354879Nunc™ CryoColor Vial Closure Coder, PSBlueCase of 2000
354879PK
also known as 354879
Nunc™ CryoColor Vial Closure Coder, PSBluePack of 500
354968Nunc™ CryoColor Vial Closure Coder, PSRedCase of 2000
354968PK
also known as 354968
Nunc™ CryoColor Vial Closure Coder, PSRedPack of 500
355018Nunc™ CryoColor Vial Closure Coder, PSGreenCase of 2000
355018PK
also known as 355018
Nunc™ CryoColor Vial Closure Coder, PSGreenPack of 500
355077Nunc™ CryoColor Vial Closure Coder, PSYellowCase of 2000
355077PK
also known as 355077
Nunc™ CryoColor Vial Closure Coder, PSYellowPack of 500
355158Nunc™ CryoColor Vial Closure Coder, PSOrangeCase of 2000
355158PK
also known as 355158
Nunc™ CryoColor Vial Closure Coder, PSOrangePack of 500
375868Nunc™ CryoColor Vial Closure Coder, PSBrownCase of 2000
375868PK
also known as 375868
Nunc™ CryoColor Vial Closure Coder, PSBrownPack of 500
375884Nunc™ CryoColor Vial Closure Coder, PSPinkCase of 2000
375884PK
also known as 375884
Nunc™ CryoColor Vial Closure Coder, PSPinkPack of 500
375906Nunc™ CryoColor Vial Closure Coder, PSGrayCase of 2000
375906PK
also known as 375906
Nunc™ CryoColor Vial Closure Coder, PSGrayPack of 500
375922Nunc™ CryoColor Vial Closure Coder, PSPurpleCase of 2000
375922PK
also known as 375922
Nunc™ CryoColor Vial Closure Coder, PSPurplePack of 500
375930Nunc™ CryoColor Vial Closure Coder, PSAssortedCase of 2000
375930PK
also known as 375930
Nunc™ CryoColor Vial Closure Coder, PSAssortedPack of 500
5045-0003Nalgene™ Color coderBlueCase of 500
5045-0003PK
also known as 5045-0003
Nalgene™ Color coderBluePack of 100
5045-0000Nalgene™ Color coderWhiteCase of 500
5045-0000PK
also known as 5045-0000
Nalgene™ Color coderWhitePack of 100
5045-0002Nalgene™ Color coderYellowCase of 500
5045-0002PK
also known as 5045-0002
Nalgene™ Color coderYellowPack of 100
5045-0004Nalgene™ Color coderGreenCase of 500
5045-0004PK
also known as 5045-0004
Nalgene™ Color coderGreenPack of 100
5045-0005PK
also known as 5045-0005
Nalgene™ Color coderRedPack of 100
Catalog number 5045-0005
Price (USD)/ Case of 500
107.35
Online exclusive
113.00 
Save 5.65 (5%)
-
Add to cart
Description:
Nalgene™ Color coder
Color:
Red
Unit Size:
Case of 500
Price (USD)/ Case of 500
107.35
Online exclusive
113.00 
Save 5.65 (5%)
Add to cart
Cryo Vial Closure Color Coders
Catalog number5045-0005
Price (USD)/ Case of 500
107.35
Online exclusive
113.00 
Save 5.65 (5%)
-
Add to cart
Make it possible to identify specific vials quickly and simplify your inventory using color coders that fit into the top of Nalgene™ Cryogenic vial closures or Nunc™ Cryogenic Tube closures.
  • Nalgene™ color coders are made from polystyrene and fit only in Nalgene™ cryogenic vial closures. Available in 5 colors
  • Nunc™ color coders are made from polycarbonate and fit only in Nunc™ cryogenic tube closures and Cryobank 2ml and 5mL tube caps. Available in 10 colors or as an assortment.
Specifications
DescriptionNalgene™ Color coder
ColorRed
MaterialPolycarbonate
Product LineNalgene
Quantity100/Pk., 500/Cs.
TypeVial Closure
Unit SizeCase of 500