Barnstead™ Point of Use Water Purification Systems Ultrafilters
Barnstead™ Point of Use Water Purification Systems Ultrafilters
Thermo Scientific™
Barnstead™ Point of Use Water Purification Systems Ultrafilters
Lengthen the life of your water purification system with these replacement ultrafilters, which are automatically flushed.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)
50133981MicroPure, Smart2Pure (UF and UF/UV units only)
50133980GenPure, GenPure Pro, GenPure xCAD
Catalog number 50133981
Price (USD)/ Each
604.70
Online exclusive
708.00 
Save 103.30 (15%)
-
Add to cart
For Use With (Equipment):
MicroPure, Smart2Pure (UF and UF/UV units only)
Price (USD)/ Each
604.70
Online exclusive
708.00 
Save 103.30 (15%)
Add to cart
Barnstead™ Point of Use Water Purification Systems Ultrafilters
Catalog number50133981
Price (USD)/ Each
604.70
Online exclusive
708.00 
Save 103.30 (15%)
-
Add to cart
Lengthen the life of your water purification system with replacement Thermo Scientific™ Barnstead™ Point-of-Use Water Purification Systems Ultrafilters. For use with Barnstead point-of-use water purification systems, these internal ultrafilters are flushed automatically to allow for a long two-year lifetime.
Use with the following Barnstead point-of-use water purification systems:
Part 50133980:
  • GenPure™†
  • GenPure™ Pro†
  • GenPure™ xCAD†
†UF, UF/UV and UF/UV-TOC units only
Part 50133981:
  • MicroPure™‡
  • Smart2 Pure™‡
‡UF and UF/UV units only
Specifications
DescriptionUltrafilter; For use with MicroPure, Smart2 Pure (UF and UF/UV units only)
For Use With (Equipment)MicroPure, Smart2Pure (UF and UF/UV units only)
TypeUltrafilter
Unit SizeEach