Catalog Number | Description |
---|---|
50088133 | Compact Stirrer w/Telemodul 20 C; 100-240V; 3/6/9/12W |
50088122 | Maxi Stirrer w/Telemodul 40 C; |
50088135 | Maxi Stirrer w/Telemodul 20 C; 100-240V |
50088143 | Maxi Stirrer w/Standard Telemodul; 230V Euro |
50088145 | Maxi Stirrer w/Standard Telemodul; 230V UK |
50088146 | Maxi Stirrer w/Standard Telemodul; 110V Japan |
50088147 | Maxi Stirrer w/Standard Telemodul; 115V US |
50088152 | Compact Stirrer w/Standard Telemodul; 230V Euro |
50088153 | Compact Stirrer w/Standard Telemodul; 230V UK |
50088154 | Compact Stirrer w/Standard Telemodul; 100V Japan |
50094111 | Compact Stirrer w/Standard Telemodul; 230V Australia |
Handle challenging stirring environments with Thermo Scientific™ Cimarec™ i Compact and Maxi Stirrers, which are 100% maintenance- and wear-free, hermetically sealed units. With powerful magnets and no moving parts, these inductive-drive stirrers are designed to go into water baths and incubators. The Cimarec™ i Compact and Maxi series stirrers come standard with your choice of controller.
Maxi Stirrer with External Control:
Compact Stirrer with External Control:
Ordering Information:
Choose from three different control units (included with stirrer). Choose the standard Telemodul controller with its space-saving design, the Telemodul 20 C with microprocessor controls, adjustable power setting and programmability or the Telemodul 40 C with higher speeds, more power settings and PC control. Stirrers are also available without controller, for use with benchtop distribution system.
Includes:
100-240V units contain a cord set with various plugs.
Warranty: 5 years