Immuno Clear Standard Modules
Immuno Clear Standard Modules
Thermo Scientific™

Immuno Clear Standard Modules

Perform applications that deliver well-to-well reproducibility with these immuno clear standard modules. Manufactured to fit standard equipment.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberWell DesignSurface TreatmentVolume (Metric) WorkingWell Shape
473709C12MaxiSorp250 μLRound, C-bottom
434797F8MaxiSorp350 μLRound, Flat Bottom (F-well)
441653C8MaxiSorp250 μLRound, C-bottom
444865C8PolySorp250 μLRound, C-bottom
445101C8MaxiSorp250 μLRound, C-bottom
467120F8MediSorp350 μLRound, Flat Bottom (F-well)
467140F8MultiSorp350 μLRound, Flat Bottom (F-well)
467466F16 (2x8 wells)MaxiSorp350 μLRound, Flat Bottom (F-well)
467679F16PolySorp350 μLRound, Flat Bottom (F-well)
468667F8MaxiSorp350 μLRound, Flat Bottom (F-well)
469078F8PolySorp350 μLRound, Flat Bottom (F-well)
469264U16Hydrophilic250 μLRound
469914F16 (2x8 wells)MaxiSorp350 μLRound, Flat Bottom (F-well)
469922F16 (2x8 wells)PolySorp350 μLRound, Flat Bottom (F-well)
469949F8MaxiSorp350 μLRound, Flat Bottom (F-well)
469957F8PolySorp350 μLRound, Flat Bottom (F-well)
473717C12PolySorp250 μLRound, C-bottom
475078U8MaxiSorp250 μLRound
475086U8PolySorp250 μLRound
6302F12Immulon 2 HB330 μLRound, Flat Bottom (F-well)
6309F12Immulon 2 HB330 μLRound, Flat Bottom (F-well)
6310F12Immulon 1 B330 μLRound, Flat Bottom (F-well)
6405F12Immulon 4 HBX330 μLRound, Flat Bottom (F-well)
95029100F8Enhanced Binding350 μLRound, Flat Bottom (F-well)
6301TS1
also known as 6301
F12Immulon 1 B330 μLRound, Flat Bottom (F-well)
Catalog number 473709
Price (USD)/ Case of 60
592.20
Online exclusive
658.00 
Save 65.80 (10%)
-
Add to cart
Well Design:
C12
Surface Treatment:
MaxiSorp
Volume (Metric) Working:
250 μL
Well Shape:
Round, C-bottom
Price (USD)/ Case of 60
592.20
Online exclusive
658.00 
Save 65.80 (10%)
Add to cart
Immuno Clear Standard Modules
Catalog number473709
Price (USD)/ Case of 60
592.20
Online exclusive
658.00 
Save 65.80 (10%)
-
Add to cart

Perform applications which are delivering well-to-well reproducibility with Thermo Scientific™ Immuno Clear Standard Modules, which feature Nunc™, Immulon™ and Microtiter™ module plates developed for ELISA techniques. Manufactured to fit standard equipment, modules provide flexible use of components with small sample sizes. They are backed by industry-leading quality standards and certifications.

Features:

  • Several strip formats: Eight-well strip, 12-well strip, 16-well strip (two columns of eight wells)
  • Several well shapes: flat bottom, C bottom (radius edge), U shape wells, StarWells (wells have fins to increase the surface area and enhance performance)
  • Several surfaces to maximize assay performance
  • Modules that remain in frame when inverted
  • Stackable frame marked for lid orientation
  • Polystyrene wells in polyethylene frame
  • Ext. Dim.: 128 × 86mm

Choice of Surface:

  • Thermo Scientific™ PolySorp™, Immulon™ 1B, and Universal Binding (UB) are for adsorption of hydrophobic molecules.
  • Thermo Scientific™ Medisorp™ and Immulon 2HB are slightly hydrophilic and bind a diverse range of biomolecules.
  • Thermo Scientific™ MaxiSorp™, Immulon 4 HBX, and Enhanced Binding(EB) are hydrophilic and ideal for antibody sandwich assays.
  • Thermo Scientific™ MultiSorp™ are very hydrophilic and works well for glycans and water soluble proteins.

Ordering Information:

Modules are freestanding. Cat. No. 95029440 and 464394 are certified. Frames and modules are available separately. Sealing tape and strip caps are available. Color-coded modules are available on request. Contact your customer service or sales representative for further details.

WARNING: Cancer – www.P65Warnings.ca.gov
Specifications
BreakapartNo
Binding PropertyHydrophilic
Certifications/ComplianceCertified for adsorption of IgG; CV of <5% between wells
CertifiedYes
FormatStandard
Detection MethodColorimetric
Array8 x 12
Packaging60 Cases
ColorClear
Product TypeFrame
MaterialPolystyrene
Volume (Metric) Well350 μL
No. of Wells96
Volume (Metric) Working250 μL
Surface TreatmentMaxiSorp
Well ShapeRound, C-bottom
No. per Case60
No. per Pack10
Plate BlockingNone
Quantity60/Cs.
Target MoleculeAntibodies (Glycoproteins)
Well DesignC12
Unit SizeCase of 60