E1-ClipTip™ Bluetooth™ Electronic Multichannel Pipettes
E1-ClipTip™ Bluetooth™ Electronic Multichannel Pipettes
Thermo Scientific™

E1-ClipTip™ Bluetooth™ Electronic Multichannel Pipettes

Automate daily 96-and 384-well microplate pipetting tasks with the sealed-in security of ClipTip technology in these electronic multichannel pipettes, which offer both versatility and customization.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNumber of ChannelsVolume (Metric)Compatible TipsType
4671010BT120.5 to 12.5 μLClipTip 12.5Electronic Pipette
4671000BT80.5 to 12.5 μLClipTip 12.5 Electronic Pipette
4671040BT82 to 125 μLClipTip 200Electronic Pipette
4671070BT810 to 300 μLClipTip 300Electronic Pipette
4671100BT815 to 1250 μLClipTip 1250Electronic Pipette
4671050BT122 to 125 μLClipTip 200Electronic Pipette
4671080BT1210 to 300 μLClipTip 300Electronic Pipette
4671090BT1230 to 850 μLClipTip 1000Electronic Pipette
4671020BT160.5 to 12.5 μLClipTip 384 12.5Electronic Pipette
4671030BT161 to 30 μLClipTip 384 30Electronic Pipette
4671060BT162 to 125 μLClipTip 384 125Electronic Pipette
Catalog number 4671010BT
Price (USD)/ Each
1,946.55
Online exclusive
2049.00 
Save 102.45 (5%)
-
Add to cart
Number of Channels:
12
Volume (Metric):
0.5 to 12.5 μL
Compatible Tips:
ClipTip 12.5
Type:
Electronic Pipette
Price (USD)/ Each
1,946.55
Online exclusive
2049.00 
Save 102.45 (5%)
Add to cart
E1-ClipTip™ Bluetooth™ Electronic Multichannel Pipettes
Catalog number4671010BT
Price (USD)/ Each
1,946.55
Online exclusive
2049.00 
Save 102.45 (5%)
-
Add to cart

With enhanced versatility and customization, the Thermo Scientific™ E1-ClipTip™ Electronic Multichannel Pipette automates daily 96-and 384-well microplate pipetting tasks with the sealed-in security of Thermo Scientific™ ClipTip™ technology. Enjoy pipetting again with optimum functionality for every application and user preference. The NEW My Pipette™ Creator app enables programming of E1-ClipTip from the comfort of your PC.

Innovative ClipTip interlocking tip interface technology*
  • Tip is locked and sealed in place until ejected — no dropped tips that waste valuable samples
  • Complete seal to secure the sample volume in each tip for enhanced accuracy and precision
  • Tips are attached and ejected with a light touch — eliminates the frustration of banging tips on the pipette

My Pipette Creator to revolutionize the way you pipette with a new era of technology
  • New web-based pipetting app powered by Thermo Fisher Cloud
  • Enables efficient, centralized programming and sharing of protocols between E1-ClipTip pipettes and colleagues
  • Offers pre-programmed protocols, to download and share, for many of your favorite reagent kits and pipetting protocols
  • Connect E1-ClipTip pipette to the app via a Bluetooth** or USB connection.
  • Accelerate collaboration and reproducibility, all from the comfort of your computer.

E1-ClipTip pipette to help you enjoy pipetting
  • E1-ClipTip is designed for optimum comfort and ease of use: the electronic tip ejection and index finger pipetting action let your thumb relax during pipetting.
  • Two operating options: Step-based Matrix or Presets with settings for your most common applications.
  • The Programs function allows you to store 30 your most common Matrix or Presets protocols with a specific name.
  • Connect your E1-ClipTip pipette to My Pipette Creator app conveniently via Bluetooth**
  • Five different user profiles offer customized settings for multiple users and protocols
  • MyPipette™ function to customize your main menu view by organizing the icons in your preferred order or creating shortcuts for your favorite programs and pipetting functions. You can also protect important settings from unintentional editing with a password.
  • Service and calibration trackers remind you when the pipette needs to be sent for service
  • Li-ion battery for long lasting performance and fast recharging. Recharging with a charging stand or a plug, you decide.
  • 2-year warranty with convenient web registration

E1-ClipTip pipette and ClipTip tips for every application
  • Choose your E1-ClipTip, volume range from 0.5μL to 1250μL:
    • Single channels
    • Multichannels with 8, 12 or 16 channels in 96 and 384 formats
    • Adjustable tip spacing multichannels with 6, 8, 12 channels in 96 and 384 formats
  • Wide selection of ClipTip tips:
    • ClipTip 96-format tips are available in 20, 50, 200, 300, 1000 and 1250μl
    • ClipTip 384-format tips are available in 12.5, 30 and 125μl
    • Several packaging options: filtered, sterile, non-sterile, racked and reload


* E1-ClipTip 12.5 and all 384 E1-ClipTip pipettes utilize snap and seal technology.
** Bluetooth models available in following countries/regions: Andorra, Australia, Austria, Belgium, Bulgaria, Canada, China, Croatia, Cyprus, Czech Republic, Denmark, Estonia, Finland, France, Germany, Greece, Hong Kong, Hungary, Iceland, India, Ireland, Italy, Japan, Korea (Rep. of), Latvia, Liechtenstein, Lithuania, Luxembourg, Malta, Monaco, Netherlands, New Zealand, Norway, Poland, Portugal, Romania, San Marino, Singapore, Slovakia, Slovenia, Spain, Sweden, Switzerland, Taiwan, United Kingdom, United States and Vatican. For other countries catalog numbers without 'BT' extension should be used.




All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified. The Bluetooth™ is a trademark of Bluetooth SIG, Inc.
Specifications
Compatible TipsClipTip 12.5
ColorPink
For Use WithClipTip™ 12.5
ModelBluetooth
Number of Channels12
Product LineE1-ClipTip™
ProgrammabilityYes
TypeElectronic Pipette
Volume (Metric)0.5 to 12.5 μL
Increments0.01μL
Unit SizeEach