Finnpipette™ F2 Multichannel Pipettes
Finnpipette™ F2 Multichannel Pipettes
Thermo Scientific™
Finnpipette™ F2 Multichannel Pipettes
Use this multichannel pipette for highly demanding applications where robustness and durability are essential.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNumber of ChannelsVolume (Metric)Compatible Tips
4662080161 to 10 μLFinntip 20 Micro, Filter 20 Micro
466200081 to 10 μLFinntip 10 Micro, Filter 10 Micro, Flex 10, Flex Filter 10
466201085 to 50 μLFinntip 250, Filter 100, Flex 200, Flex Filter 100, 200 Ext, Filter 100 Ext
4662020810 to 100 μLFinntip 250, Filter 100, Flex 200, Flex Filter 100, 200 Ext, Filter 100 Ext, 250 Wide
4662030830 to 300 μLFinntip 300, Filter 300, Flex 300, Flex Filter 300
4662040121 to 10 μLFinntip 10 Micro, Filter 10 Micro, Flex 10, Flex Filter 10
4662050125 to 50 μLFinntip 250, Filter 100, Flex 200, Flex Filter 100, 200 Ext, Filter 100 Ext
46620601210 to 100 μLFinntip 250, Filter 100, Flex 200, Flex Filter 100, 200 Ext, Filter 100 Ext, 250 Wide
46620701230 to 300 μLFinntip 300, Filter 300, Flex 300, Flex Filter 300
4662090165 to 50 μLFinntip 50 Micro, Filter 50 Micro
Catalog number 4662080
Price (USD)/ Each
1,276.20
Online exclusive
1418.00 
Save 141.80 (10%)
-
Add to cart
Number of Channels:
16
Volume (Metric):
1 to 10 μL
Compatible Tips:
Finntip 20 Micro, Filter 20 Micro
Price (USD)/ Each
1,276.20
Online exclusive
1418.00 
Save 141.80 (10%)
Add to cart
Finnpipette™ F2 Multichannel Pipettes
Catalog number4662080
Price (USD)/ Each
1,276.20
Online exclusive
1418.00 
Save 141.80 (10%)
-
Add to cart
Use the Thermo Scientific™ Finnpipette™ F2 Multichannel Pipette for highly demanding applications where robustness and durability are essential. With a sleek, rugged design, this pipette is built with PVDF components that stand up to harsh chemicals and the damaging effects of UV light. Fully autoclavable and available in 8-, 12- and 16-channel models with a selection of volume ranges. In the low volume models, the super blow-out function ensures accurate dispensing for even the lowest volumes.
For decades, Thermo Scientific Finnpipette pipetting systems have delivered unmatched productivity and ergonomics across thousands of labs and applications worldwide.
Finnpipette F2: Proven reliable for long-term intensive use
Rugged and Durable
Finnpipette F2 contains tough PVDF components that stand up to harsh chemicals and the damaging effects of UV light. Its rugged design withstands physical use without damage.
Fully Autoclavable
Decontamination of the Finnpipette F2 is simple as there is no need to disassemble the F2 for autoclaving, minimizing disruption and downtime.
Maintenance is Easy
Simply detach the tip cone for efficient daily maintenance or decontamination when not using an autoclave.
Super Blow-out
150% increase in air boost to ensure efficient delivery of micro-volumes and prevent capillary action in 50μL models and below.
Wide Selection of Finntip Pipette Tips
Finntips are designed and manufactured expressly for Finnpipette models, enabling optimal performance, precision and accuracy.
5 Year Warranty
With convenient online registration, and may vary by country.
Specifications
Compatible TipsFinntip 20 Micro, Filter 20 Micro
DescriptionFinnpipette™ F2
AutoclavableAutoclavable
ColorPurple
Number of Channels16
Product LineFinnpipette™ F2
TypePipette
Volume (Metric)1 to 10 μL
Increments0.02μL
Color CodePurple
Unit SizeEach