Specifications
Format96-well Plate
Isolation TechnologyEnzymatic, Magnetic Bead
Sample TypeBacteria (Gram +/-), Bacterial Culture, Food and Environmental, Liquid Samples (e.g. Serum), Water
Final Product TypeBacterial DNA, Viral RNA
For Use With (Equipment)MagMAX™ Express-96 Deep Well Magnetic Particle Processor
For Use With (Application)Real-Time Quantitative PCR (qPCR), PCR
High-throughput CompatibilityHigh-throughput Compatible, Automated Protocols
No. of Reactions100 Preps
Product LinePrepSEQ™
TargetGenomic DNA, Viral RNA
Starting Material Amount≤1 mL culture
Unit Size100 preps