You need to enable JavaScript to run this app.
ViiA™ 7, QuantStudio™ 6 Flex, QuantStudio™ 7 Flex and QuantStudio™ 12k Flex System Block Upgrade Kit, Fast, 96 well
Applied Biosystems™
ViiA™ 7, QuantStudio™ 6 Flex, QuantStudio™ 7 Flex and QuantStudio™ 12k Flex System Block Upgrade Kit, Fast, 96 well
This kit is an upgrade that allows the following systems to operate using a Fast, 96 well block.ViiA™ 7
Real-Time
Read more
Have Questions?
Catalog number
4453544
Price (USD)
/ 1 upgrade
-
Request A Quote
This kit is an upgrade that allows the following systems to operate using a Fast, 96 well block.
ViiA™ 7 Real-Time PCR System
QuantStudio™ 6 Flex Real-Time PCR System
QuantStudio™ 7 Flex Real-Time PCR System
QuantStudio™ 12K Flex Real-Time PCR System
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Format
96-well Plate
For Use With (Equipment)
QuantStudio™
Reaction Speed
Fast
Capacity
1 x 96-well Plate
Product Line
ViiA™
Product Type
System Block Upgrade Kit
Description
Fast 96-well Plate
Quantity
1 upgrade
Unit Size
1 upgrade
Contents & Storage
Fast 96-well Block (0.1mL), heated cover, plate adaptor and calibration kit.