Specifications
Certifications/ComplianceTitan3 Polypropylene Syringe Filter Housings Meet The Requirements of 21 CFR 177.1520. Membrane: HPLC Certified PTFE, with Polypropylene Support .
ColorTransparent Yellow
DisposableSingle-use
Material (Membrane)PTFE (Hydrophobic)
Pore Size0.45 μm
SterilityNon-sterile
For Use With (Application)Sample Filtration: aggressive organic, highly basic, or hot solutions, aqueous solutions without prewetting with an alcohol
Hold up Volume (After Air Purge)<157 μL
Housing MaterialPolypropylene
PrefilterYes
Pressure Rating210 psi
Product LineTitan3
Quantity100/Pk.
TypeSyringe Filter
Volume (Metric)<137 μL (Hold Up)
Diameter (Metric)30 mm
Diameter (Metric) Membrane30 mm
Unit SizePack of 100