Megaplex™ Primer Pools, Rodent Pools B v3.0
Product Image
Applied Biosystems™

Megaplex™ Primer Pools, Rodent Pools B v3.0

Megaplex™ Primer Pools, Rodent Pool B v3.0 is available under a single part number containing both the Megaplex™ RT PrimersRead more
Have Questions?
Catalog number 4444752
Price (USD)/ 50 reactions
1,606.00
-
Add to cart
Price (USD)/ 50 reactions
1,606.00
Add to cart
Megaplex™ Primer Pools, Rodent Pools B v3.0
Catalog number4444752
Price (USD)/ 50 reactions
1,606.00
-
Add to cart
Megaplex™ Primer Pools, Rodent Pool B v3.0 is available under a single part number containing both the Megaplex™ RT Primers and Megaplex™ PreAmp Primers, Rodent Pools B v3.0. Megaplex™ RT Primers, Rodent Pool B contains RT primers for 306 and 135 unique microRNAs for mouse and rat respectively plus 4 species specific controls. For use when assay sensitivity is of the utmost importance, Megaplex™ PreAmp Primers significantly enhance the ability to detect low expressed microRNAs enabling the generation of a comprehensive expression profile using far less sample, as low as 1 ng of input total RNA. The microRNA targets represented in this pool tend be less well characterized, more narrowly expressed and⁄or expressed at a lower level compared to Rodent Pool A. Rodent Pool B is intended for use with the TaqMan Rodent MicroRNA B Array v3.0, however, it can also be used to prepare cDNA for real-time analysis using the individual TaqMan® MicroRNA Assays.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
GC-Rich PCR PerformanceLow
PCR MethodqPCR
PrimerRT Primers, PreAmp Primers
Product LineMegaplex™, TaqMan™
Product TypePrimer Pool
Quantity50 reactions
SpeciesRat, Mouse
Sufficient For50 Reactions
For Use With (Application)miRNA analysis
No. of Reactions50 Reactions
Unit Size50 reactions
Contents & Storage
Store at -20°C.