OpenArray™ Plate Compression Pad for Dual Flat Block GeneAmp™ PCR System 9700
OpenArray™ Plate Compression Pad for Dual Flat Block GeneAmp™ PCR System 9700
Applied Biosystems™
OpenArray™ Plate Compression Pad for Dual Flat Block GeneAmp™ PCR System 9700
The OpenArray™ Plate Compression Pad for Dual Flat Block GeneAmp™ PCR System 9700 properly and evenly positions up to eightRead more
Have Questions?
Catalog number 4426825
Price (USD)/ 1 pad
75.00
-
Add to cart
Price (USD)/ 1 pad
75.00
Add to cart
OpenArray™ Plate Compression Pad for Dual Flat Block GeneAmp™ PCR System 9700
Catalog number4426825
Price (USD)/ 1 pad
75.00
-
Add to cart
The OpenArray™ Plate Compression Pad for Dual Flat Block GeneAmp™ PCR System 9700 properly and evenly positions up to eight TaqMan™ OpenArray™ Genotyping Plates for PCR amplification. It is intended for use with the Dual Flat Block GeneAmp™ PCR System 9700 System and OpenArray™ Plate Adaptor for Dual Flat Block GeneAmp™ PCR System 9700.
For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)GeneAmp 9700
Product LineGeneAmp™, OpenArray™
Product TypePlate Compression Pad
For Use With (Application)PCR
Shipping ConditionRoom Temperature
Quantity1 pad
Plate CompatibilityOpenArray™ Plates
Unit Size1 pad
Contents & Storage
Required compression pad for using OpenArray™ plate holder with the GeneAmp 9700 Dual flat block PCR system