PrepSEQ™ Rapid Spin Sample Preparation Kit - Extra Clean with Proteinase K
Applied Biosystems™
PrepSEQ™ Rapid Spin Sample Preparation Kit - Extra Clean with Proteinase K
The PrepSEQ™ Spin Sample Preparation Kit – Extra Clean with Proteinase K is recommended for difficult to lyse bacteria, likeRead more
Have Questions?
Catalog number 4426715
Price (USD)/ 1 kit
495.00
-
Add to cart
Price (USD)/ 1 kit
495.00
Add to cart
PrepSEQ™ Rapid Spin Sample Preparation Kit - Extra Clean with Proteinase K
Catalog number4426715
Price (USD)/ 1 kit
495.00
-
Add to cart
The PrepSEQ™ Spin Sample Preparation Kit – Extra Clean with Proteinase K is recommended for difficult to lyse bacteria, like Listeria monocytogenes, that also has a high lipid content, such as soft cheeses. The kit was developed for the rapid preparation of DNA from food and environmental samples for use in PCR amplification reactions. Using a simple spin protocol, PrepSEQ™ Rapid Spin efficiently inactivates PCR inhibitors. For laboratories involved in food and environmental testing, PCR-based methods are rapidly becoming the standard for accurate, sensitive bacterial detection. This has created the demand for a sample preparation method that is fast, cost-effective and capable of extracting high-quality DNA from a broad range of sample types. While such methods have been developed for dedicated, high-throughput laboratories, there is a growing need for methods suitable to low-to-medium throughput applications. The PrepSEQ™ Rapid Spin Kit – Extra Clean with Proteinase K meets this need. • Designed for Food Safety and Environmental Monitoring applications • Supports low-to-medium throughput work loads (up to 100 samples a day) • Inactivates PCR inhibitors so you will most likely not have to repeat the template preparation step • Uses a simple spin protocol Optimized for PCR-based Food Safety and Environmental Monitoring For laboratories involved in food safety and environmental monitoring, PCR-based methods are rapidly becoming the standard for accurate, sensitive bacterial detection. Use this sample preparation method for fast, cost-effective extraction of high-quality DNA from a broad range of sample types including chocolate, soft cheeses, and soil. Even samples with difficult-to-lyse bacteria, such as Listeria monocytogenes, produce excellent results with the addition of a Proteinase K digestion step.
Designed for Laboratories that Prepare up to 100 Samples a Day Kits exist for dedicated, high-throughput laboratories, but now you have a kit that is suitable for low-to-medium throughput applications. This kit is designed for laboratories that routinely prepare up to 100 samples a day.
Removes PCR Inhibitors The PrepSEQ™ Rapid Spin Sample Preparation Kit offers a simple, cost-effective way to prepare bacterial DNA from broth cultures. During the extraction process, the media inside the spin column removes PCR inhibitors from the sample, allowing the bacteria to pass through into the sample collection tube where it can be stored or used for PCR.
. For Research Use Only. Not for use in diagnostics procedures.
Each kit contains 100 spin columns, 200 microcentrifuge tubes, Lysis Buffer (store at 4°C), and Proteinase K (store at -20°C). 100 samples can be prepared using this kit.