TaqMan™ Array, Human IL-2 Active & Quiescent T Cells, Fast 96-well
Not what you're looking for? Search all of our TaqMan® arrays for your gene or pathway of interest.
Product Image
Applied Biosystems™

TaqMan™ Array, Human IL-2 Active & Quiescent T Cells, Fast 96-well

These 96-well plates are pre-configured with the most appropriate TaqMan® Gene Expression Assays for a specific biological process, pathway, orRead more
Have Questions?
Catalog number 4418789
Price (USD)/ 1 x 96 well plate
230.00
-
Add to cart
Price (USD)/ 1 x 96 well plate
230.00
Add to cart
TaqMan™ Array, Human IL-2 Active & Quiescent T Cells, Fast 96-well
Catalog number4418789
Price (USD)/ 1 x 96 well plate
230.00
-
Add to cart
These 96-well plates are pre-configured with the most appropriate TaqMan® Gene Expression Assays for a specific biological process, pathway, or disease state. Each plate contains predefined assays and endogenous controls dried-down in the wells, ready for accurate assessment of an entire gene signature in one simple experiment. The TaqMan® Array Human IL-2 Gene Expression In Activated And Quiescent T-Cells 96-well Plate contains 28 assays to IL-2 gene expression in activated and quiescent T-Cells associated genes and 4 assays to candidate endogenous control genes. All assays are plated in triplicate.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
TypeArray Plate
Format96-array Plate
SpeciesHuman
FormDried Down
IncludesTaqMan Assay for One 10 uL Reaction
Internal Probe ModificationMGB (Minor Groove Binder) (3'), Non-Fluorescent Quencher (3'), FAM (5')
Label or DyeFAM
Product LineTaqMan™
Quantity1 x 96-well plate
Reaction SpeedFast
Research CategoryBiomarker-Related Pathway(s), Signal Transduction
Unit Size1 x 96 well plate
Contents & Storage
Each TaqMan™ Array Plate well contains a single TaqMan™ Assay for one 10 μL reaction. The plates can be stored at room temperature or at 4°C.