TaqMan™ Array, Human NFKB, Fast 96-well
Not what you're looking for? Search all of our TaqMan® arrays for your gene or pathway of interest.
Do you need this panel in a 384-well plate? Please visit Specialty Plating to learn more.
Product Image
Applied Biosystems™

TaqMan™ Array, Human NFKB, Fast 96-well

These 96-well plates are pre-configured with the most appropriate TaqMan® Gene Expression Assays for a specific biological process, pathway, orRead more
Have Questions?
Catalog number 4418740
Price (USD)/ 1 x 96 well plate
230.00
-
Add to cart
Price (USD)/ 1 x 96 well plate
230.00
Add to cart
TaqMan™ Array, Human NFKB, Fast 96-well
Catalog number4418740
Price (USD)/ 1 x 96 well plate
230.00
-
Add to cart
These 96-well plates are pre-configured with the most appropriate TaqMan® Gene Expression Assays for a specific biological process, pathway, or disease state. Each plate contains predefined assays and endogenous controls dried-down in the wells, ready for accurate assessment of an entire gene signature in one simple experiment. The panel of assays in the TaqMan® Array 96-well Human NFkB Pathway Plate targets genes which encode the 5 proteins of the Rel⁄NFKB family: NFKB1, NFKB2, REL A, REL B, and REL. Also on the panel are genes from families that include: the IkB kinase; IkBs (inhibitor of kB); Toll-like receptor (TLR); Tumor Necrosis Factor (TNF); Tumor Necrosios Factor Receptors (TNFR); and Tumor Necrosis Factor Receptor Associated Factor (TRAF). Additional genes associated with NFKB function in apoptosis, immune⁄inflammation responses, as well as chemokines and cytokines are included.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
FormDried Down
Format96-well plate
Reaction SpeedFast
Internal Probe ModificationMGB (Minor Groove Binder) (3'), Non-Fluorescent Quencher (3'), FAM (5')
Label or DyeFAM
Product LineTaqMan™
Research CategoryImmunology, Biomarker-Related Pathway(s), Extracellular Matrix & Cell Adhesion, Apoptosis
SpeciesHuman
For Use With (Application)Gene Expression
No. of Reactions1 x 10 μL Reactions
Unit Size1 x 96 well plate
Contents & Storage
Each TaqMan™ Array Plate well contains a single TaqMan™ Assay for one 10 μL reaction. The plates can be stored at room temperature or at 4°C.