OpenArray™ 384-well Sample Plates
Product Image
Applied Biosystems™

OpenArray™ 384-well Sample Plates

The OpenArray® 384-well Sample Plates are for use with the QuantStudio™ 12K Flex Real-Time PCR System. They are low volumeRead more
Have Questions?
Catalog number 4406947
Price (USD)/ 10 plates
151.00
-
Add to cart
Price (USD)/ 10 plates
151.00
Add to cart
OpenArray™ 384-well Sample Plates
Catalog number4406947
Price (USD)/ 10 plates
151.00
-
Add to cart
The OpenArray® 384-well Sample Plates are for use with the QuantStudio™ 12K Flex Real-Time PCR System. They are low volume microplates used in the QuantStudio™ OpenArray® Accufill™ System for sample loading onto OpenArray® plates.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Format384-well Plate
For Use With (Equipment)QuantStudio™ 12 K Flex
Reaction SpeedStandard
Product LineOpenArray™
Product TypeSample Plate
For Use With (Application)Real Time PCR (qPCR)
Shipping ConditionRoom Temperature
DescriptionClear
Quantity10 plates
BarcodeNo Barcode
ColorClear
Unit Size10 plates
Contents & Storage
Each box contains 10 x 384-well plates, enough for 80 OpenArray™ Real-Time PCR plates.