3500 Genetic Analyzer for Human Identification
3500 Genetic Analyzer for Human Identification
Applied Biosystems™

3500 Genetic Analyzer for Human Identification

The 8-capillary 3500 Genetic Analyzer sets the standard in capillary electrophoresis. The 3500-series instruments are the first genetic analyzers designedRead more
Have Questions?
Catalog number 4406017
Price (USD)/ 1 system
-
Request A Quote
The 8-capillary 3500 Genetic Analyzer sets the standard in capillary electrophoresis. The 3500-series instruments are the first genetic analyzers designed with a specific feature set and workflow for the Human Identification application. The complete product combines the instrument with Applied Biosystems™ reagents, consumables and software, as well as industry leading support, to provide an integrated HID system solution that significantly improves ease-of-use and application efficiencies.

• Increased Consistency – Reduced signal variation with factory standardization and optional in-lane normalization
• Real-Time Data Quality Assessment – On instrument data evaluation features provide rapid identification and re-injection of samples requiring additional processing
• Improved Thermal Control – Smaller oven design and improved door seal for more consistent data migration
• HID Specific Workflow – Pre-configured for Applied Biosystems™ amplification kits to simplify run setup and software navigation
• Effortless Consumable Installation – Convenient one-time use consumable packaging
• Consumable Tracking with RFID Technology – On-instrument, real-time monitoring of consumable consumption and expiration
• Enhanced Throughput – Faster data generation with 8 or 24 capillary arrays and shorter run times
• Improved Polymer Pump Design – Reduced waste and fewer bubbles for better data quality
• Simplified Instrument Installation & Operation – New solid-state laser eliminates need for high voltage wiring and external venting
• Streamlined Instrument Maintenance – HID specific performance check and maintenance scheduling tools
• Data Chain of Custody – Controlled access and tracking through Security, Audit and eSig features

For more information, please contact your local Sales Representative.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
TypeGenetic Analyzer
For Use With (Equipment)3500 Genetic Analyzer
Analysis Time30 to 140 min. (varies depending on run module)
ThroughputRange: 80 to 368 sequencing samples per day (varies depending on run module), ≥312 samples per day (HID36 POP4 Module), ≥ 312 samples/day (HID36_POP4), ≥88 samples per day (standard sequencing with POP to 7™ and 50 cm capillaries)
Capillary Length50 cm, 36 cm
Format96-well plate, 8-well strips
Number of Capillaries8 Capillaries
Read Length≥420 bp using a 36 cm capillary and POP-4™, >850 bp using a 50 cm capillary and POP-7™ polymer
Quantity1 system
PolymerPOP-4™, POP-6™, POP-7™
Polymer ConsumptionReplace at earliest of 14 days⁄96 samples⁄12 injections (96-sample pouches), Replace at earliest of 14 days⁄384 samples⁄60 Injections (384-sample pouches), Replace at earliest of 14 days⁄960 samples⁄120 Injections (960-sample pouches)
Warranty1 Year Limited Warranty on Parts and Labor, Service Installation and Application Training Included
Dimensions (D x W x H)Electrophoresis Unit Width (Closed Door): 61 cm, Electrophoresis Unit: 122 cm (W) (open door) x 61 cm (D) x 72 cm (H)
Amperage15 A
VoltageElectrophoresis Voltage - Up to 20 kV
External ComputerHard Drive: 2x 500GB SATA 3.0 Gb/s and 8 MB Data Burst Cache, Memory: 16 GB (2x 8GB) 1600 MHz DDR3 Non-ECC. 4th Gen Intel Core I7 Processor- 3.1 GHz Turbo Processor, Operating System: Windows 7 SP1
Laser FeaturesLong-life single-line 505 nm solid-state laser excitation source., Long-life single-line 505 nm solid-state laser
Operating EnvironmentHumidity: 20%-80% (non-condensing), Room temp.: <2° fluctuation during runs, Room temp: <2° fluctuation during runs, Temperature: 15°C-30°C, Humidity: 20–80% (non-condensing)
Operating SystemWindows 7 (Service Pack 1)
SoftwareGeneMapper™ ID-X Software
Unit Size1 system
Contents & Storage
The 3500 analyzer is shipped with the following system components:

• Capillary Electrophoresis instrument.
• 3500 (8-capillary) array and POP™ polymer
• Human identification reagents and other consumables for system qualification.
• Dell™ computer workstation with flat-screen monitor.
• Integrated software for instrument control, data collection, quality control, and basecalling or sizing of samples.