TaqMan™ Array Human Stem Cell Pluripotency Panel
Not what you're looking for? Search all of our TaqMan® arrays for your gene or pathway of interest.
Do you need this panel in a 384-well plate? Please visit Specialty Plating to learn more.
Product Image
Applied Biosystems™

TaqMan™ Array Human Stem Cell Pluripotency Panel

The Applied Biosystems™ TaqMan™ Human Stem Cell Pluripotency Array is an efficient, easy-to-use micro fluidic card for the characterization ofRead more
Have Questions?
Catalog number 4385344
Price (USD)/ 4 cards
2,155.00
-
Add to cart
Price (USD)/ 4 cards
2,155.00
Add to cart
TaqMan™ Array Human Stem Cell Pluripotency Panel
Catalog number4385344
Price (USD)/ 4 cards
2,155.00
-
Add to cart

The Applied Biosystems™ TaqMan™ Human Stem Cell Pluripotency Array is an efficient, easy-to-use micro fluidic card for the characterization of undifferentiated stem cells or their differentiated derivatives. This gene signature panel contains 90 well-defined genes validated as markers for pluripotency plus 6 endogenous controls. The format of the TaqMan Array allows for 1-4 replicates to be run in parallel per array. This product is sold in packages of 4 cards.

The Human Stem Cell Pluripotency Array contains a well-defined set of validated gene expression markers to characterize human embryonic stem (ES) cell identity and assess phenotypic variations between embryonic stem cell isolates. The gene content on this TaqMan Array resulted directly from the work of the I.S.C.I consortium and characterization of human embryonic stem cell lines by the International Stem Cell Initiative.

The array is pre-formatted and inventoried, cost-effective, convenient and easy to use without needing expensive robotics. Results are reproducible and consistent across samples, studies, and labs – the same data quality from card-to-card and lot-to-lot – even with different operators.

The Right Targets

These TaqMan Arrays use Applied Biosystem's proprietary micro fluidics technology to simultaneously analyze gene expression levels by quantitative PCR for 92 test genes and 6 endogenous controls. Genes were included based on the following criteria:

  1. Expression in human embryonal carcinoma cells (hEC), primordial germ cells or human ES (hES) cells
  2. Known role in maintenance of pluripotency in hES cells
  3. Identification in microarray-based experiments as being correlated with the stem cell state
  4. Expression known to be characteristic of specific differentiation lineages in the post implantation embryo and mature tissues. These are the right targets to characterize embryonic stem cells.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
FormDried Down
Format384-well microfluidic card
Reaction SpeedStandard
Internal Probe ModificationMGB (Minor Groove Binder) (3'), Non-Fluorescent Quencher (3'), FAM (5')
Label or DyeFAM
Product LineTaqMan™
Quantity4 cards
Research CategoryStem Cell Research, Reproduction, Development
SpeciesHuman
Target90 well-defined genes validated as markers for stem cell pluripotency
For Use With (Application)Stem Cell Analysis
No. of Reactions1 x 2 μL Reactions
Unit Size4 cards
Contents & Storage
Each TaqMan™ Array well contains a single TaqMan™ Assay for one 2 μL reaction. The cards should be stored at 4°C.