7500 Real-Time PCR System, laptop
7500 Real-Time PCR System, laptop
Applied Biosystems™

7500 Real-Time PCR System, laptop

The 7500 Real-Time PCR System is a powerful platform for labs requiring superior performance and maximum dye versatility. This systemRead more
Have Questions?
Catalog number 4351104
Price (USD)/ 1 system
-
Request A Quote
The 7500 Real-Time PCR System is a powerful platform for labs requiring superior performance and maximum dye versatility. This system is a sophisticated platform for users who require extended capabilities and maximum versatility. The 3rd generation platform features an innovative optical system that enhances sensitivity and lets you access a broader range of fluorophores. The variable excitation capability allows greater sensitivity for longer wavelength (red) dyes.

• Powerful, five-color platform is calibrated for the broadest range of dyes available: FAM™⁄SYBR® Green I, VIC®⁄JOE™, NED™⁄ TAMRA™⁄ Cy3™, ROX™⁄Texas Red™, and Cy5™ dyes
• Specialized optical system enables easy and accurate calibration to new dyes without requiring the addition of new filter sets
• Advanced multi-componenting algorithm minimizes spectral crosstalk – superior for multiplexing
• User-friendly software includes plate set-up wizards, multi-plate data viewing capabilities, and advanced analysis tools to make data processing simple and straightforward

NEW - 7500 Software v2.x
Now the easy-to-use StepOne software is available for both the 7500 and 7500 Fast systems with the 7500 Software v2.x upgrade. The 7500 Software v2.x incorporates your favorite StepOne Software features, such as a variety of plate setup wizards, standard curve dilution and master mix recipe calculators, QC flags, data filters, and email notification when a run is finished. The 7500 Software v2.x also includes an enhanced Gene Expression Study package and has a variety of new melting curve protocol options, including multiple peak detection, step and hold temperature control, and customizable ramp rates.

The NEW Gene Expression Study package accommodates large studies better than any other instrument software package:
• Import an unlimited number of Comparative CT (relative quantitation) files to one study
• Group samples and view data both by technical replicate group and biological replicate group
• Use any gene(s) as an endogenous control, including averaging multiple controls together
• Enter known efficiency values to be factored into the RQ results

21 CFR Part 11 Module available
The SDS v1.4 21CFRp11 Module is a powerful tool for assisting with 21CFRp11 compliance, while still offering the flexibility of user-customizable configuration settings.
• Individual user log-ins can be added for up to four user groups, each group with designated permission settings
• User-customizable permission settings include fourteen system activities, e-signature authority designation, and additional security settings to give you maximum control over your compliance efforts
• Audit trails can be enabled or disabled depending on your traceability needs
• A selection of e-signatures is available to ease e-signatures into your workflow

Supports Many Applications
Applications include gene expression analysis, pathogen quantitation, SNP genotyping, isothermal and +⁄– assays utilizing internal positive controls. To facilitate many of these applications, Applied Biosystems® provides preformulated, ready-to-use, quality-tested, TaqMan® assays for use with the 7500 system. Now you can reduce your assay optimization efforts.

Upgrade to High-Speed Thermal Cycling
An optional upgrade to the 7500 Fast System is available. This 7500 Fast System uses our master mix formulations and enables you to shorten your real-time PCR runs to as little as 30 minutes.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Format96-well Plate
Thermal Accuracy0.25°C (35°C to 95°C after 3 min.)
Sample Ramp Rate± 1.1°C⁄s
Sensitivity1 Copy of RNase P gene (Human Genomic DNA)
Temperature Range (Metric)4 to 100°C
Reaction Volume Range20–100 μL
Passive Reference DyeROX (Separate Tube), No ROX, ROX (Pre-mixed)
Dynamic Range9 Logs
Calibrated DyeSYBR™ Green I, ROX™, NED™, TAMRA™, JOE™, Texas Red™, Cy5™, VIC™, FAM™, Cy3™
High-throughput CompatibilityMultiplexing
For Use With (Equipment)7500 System
Thermal Uniformity0.50°C (after 30 s)
Reaction SpeedStandard
Throughput4 to 5-96-well Plates per 8-hr day
OpticsCCD Camera, 5 Excitation Filters, Tungsten-halogen Lamp, 5 Emission Filters
Thermal Cycling SystemPeltier-based System
DimensionsWidth 34 cm (13.39 in.), Depth 45 cm (17.72 in.), Height 49 cm (19.29 in.)
External ComputerNotebook: Intel Core 2 Duo T5500 (1.66 GHz), 8X DVD, 80 GB HD, 1.0 G DDR2-533 MHz SDRAM
Software CompatibilityPrimer Express, 21 CFR Part 11 Module (optional), 7500 System Software (RQ included)
Precision99.7% Confidence (5000 & 10000 Copies)
Voltage120 V
Electrical Requirements950 W
Peak Block Ramp Rate2.5°C⁄s
Max. Heating Rate (Thermal Block)2.5°C
Quantity1 system
Weight34 kg (75 lb.)
Unit Size1 system
Contents & Storage
Notebook Computer