MicroAmp™ Fast Optical 96-Well Reaction Plate with Barcode, 0.1 mL
Need help? Find instrument-specific tips, troubleshooting help, and technical resources within our new Real-Time PCR and Digital PCR Instruments Support Center.
Need help choosing the right PCR/qPCR plastics? Use our on-line selection tool »
MicroAmp™ Fast Optical 96-Well Reaction Plate with Barcode, 0.1 mL
Applied Biosystems™

MicroAmp™ Fast Optical 96-Well Reaction Plate with Barcode, 0.1 mL

The Applied Biosystems™ MicroAmp™ Fast Optical 96-Well Reaction Plate with Barcode is an integral component of the Applied Biosystems™ FastRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberBarcodeQuantity
4346906Includes Barcode20 Plates
4366932Includes Barcode200 Plates
Catalog number 4346906
Price (USD)/ 20 plates
134.00
Add to cart
Barcode:
Includes Barcode
Quantity:
20 Plates
Price (USD)/ 20 plates
134.00
Add to cart
MicroAmp™ Fast Optical 96-Well Reaction Plate with Barcode, 0.1 mL
Catalog number4346906
Price (USD)/ 20 plates
134.00
-
Add to cart
The Applied Biosystems™ MicroAmp™ Fast Optical 96-Well Reaction Plate with Barcode is an integral component of the Applied Biosystems™ Fast PCR System, which reduces PCR reaction time from 2 hours to as little as 25 minutes.

• Maximum thermal conductivity for precise thermal cycling
• Reproducible, specific, and sensitive PCR results in just 25 minutes
• Validated with other system components for consistent results
• Each reaction plate includes a unique serialized, eight character number label (barcode) that is user-readable and machine-readable to prevent tracking errors

Increased Thermal Contact for Faster, More Uniform Heating
The advanced design of the MicroAmp™ Fast 96-Well Reaction Plate dramatically increases its thermal contact with the Applied Biosystems™ Veriti™ 96-Well Fast Thermal Cycler. The plate also has a consistent well-to-well thickness so all of the samples cycle through the PCR process uniformly. These features reduce experimental artifacts and help ensure fast, accurate results.

Amplify in as Little as 25 Minutes
Go fast with the Applied Biosystems™ Fast System, comprising pre-mixed Fast PCR master mixes, specially designed 0.1 mL microplates and tubes, and the Veriti™ 96-Well Fast Thermal Cycler. Now, you can amplify fragments under 500 bp from genomic backgrounds in about 25 minutes and, in some cases, as little as 10 minutes. And you can amplify large fragments faster too—up to 2 kb in as little as 40–50 minutes. Or for sequencing applications, the AmpliTaq Gold™ Fast PCR Master Mix, UP, reduces your cycling time to 50 minutes and produces amplicons optimal for high-quality sequencing results.
Specifications
BarcodeIncludes Barcode
Certifications/ComplianceCertified DNA/RNase Free
Packaging20 Plates
Product LineMicroAmp
ColorOptical
Product TypeMicroplate
For Use With (Equipment)Thermal Cycler, Genetic Analyzer, DNA Analyzer
Skirt StyleSemi
MaterialPolypropylene
Volume (Metric) Well0.2 mL
No. of Wells96
Volume (Metric) Working0.1 mL
Quantity20 Plates
Reaction SpeedFast
Shipping ConditionRoom Temperature
Unit Size20 plates
Contents & Storage
Contains 20 plates.

May be stored at room temperature. Under proper storage conditions, plates may be stored indefinitely.