BloodPrep™ DNA Purification Solution
Inquire about OEM or Commercial Supply version of this product here.
BloodPrep™ DNA Purification Solution
Invitrogen™

BloodPrep™ DNA Purification Solution

The BloodPrep™ DNA Purification Solution is a cell lysis and DNA purification solution for whole blood from any animal species,Read more
Have Questions?
Catalog number 4342775
Price (USD)/ 1,000 mL
634.10
Special Offer
Online exclusive
Ends: 16-Dec-2024
746.00 
Save 111.90 (15%)
-
Add to cart
Price (USD)/ 1,000 mL
634.10
Special Offer
Online exclusive
Ends: 16-Dec-2024
746.00 
Save 111.90 (15%)
Add to cart
BloodPrep™ DNA Purification Solution
Catalog number4342775
Price (USD)/ 1,000 mL
634.10
Special Offer
Online exclusive
Ends: 16-Dec-2024
746.00 
Save 111.90 (15%)
-
Add to cart
The BloodPrep™ DNA Purification Solution is a cell lysis and DNA purification solution for whole blood from any animal species, cultured cells, and buccal swabs for use with BloodPrep™ DNA chemistry.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Nucleic Acid Purification
Product LineBloodPrep™
Quantity1000 mL
Shipping ConditionRoom Temperature
Unit Size1,000 mL
Contents & Storage
Store at room temperature.