You need to enable JavaScript to run this app.
Lock for Notebook Computers
Applied Biosystems™
Lock for Notebook Computers
This is a security device for notebook computers.For Research Use Only. Not for use in diagnostics
procedures.
Read more
Have Questions?
Catalog number
4332279
Price (USD)
/ 1 each
396.00
-
Add to cart
Price (USD)
/ 1 each
396.00
Add to cart
Lock for Notebook Computers
Catalog number
4332279
Price (USD)
/ 1 each
396.00
-
Add to cart
This is a security device for notebook computers.
For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)
Notebook Computers
Quantity
1 each
Product Type
Computer Lock
Shipping Condition
Room Temperature
Unit Size
1 each
Contents & Storage
Store at room temperature.