Single-Channel 96-well Temperature Verification Probe for PCR 9700 and 2720 Systems, 0.2 mL
Applied Biosystems™
Single-Channel 96-well Temperature Verification Probe for PCR 9700 and 2720 Systems, 0.2 mL
The Temperature Verification Kits and Probes are engineered and manufactured specifically for Applied Biosystems to validate temperature calibration and uniformityRead more
Have Questions?
Catalog number 4319086
Price (USD)/ 1 probe
2,360.00
-
Add to cart
Price (USD)/ 1 probe
2,360.00
Add to cart
Single-Channel 96-well Temperature Verification Probe for PCR 9700 and 2720 Systems, 0.2 mL
Catalog number4319086
Price (USD)/ 1 probe
2,360.00
-
Add to cart
The Temperature Verification Kits and Probes are engineered and manufactured specifically for Applied Biosystems to validate temperature calibration and uniformity in GeneAmp™ PCR and the Veriti™ Thermal Cycler instrument systems. This is the spare probe for the 0.2mL 96-Well single channel Temperature Verification Kit (PN 4317939).
• Self-contained digital thermometer and battery operated for ease of use • Interchangeable Platinum Resistant probes
Complete Offering To expand the use of an existing Temperature Verification Kits, the individual platinum resistant probes are now available in 0.02 mL, 0.1mL, 0.2 mL, for purchase.
More information can also be found at Alpha Technics (Formerly Alpha Sensors, Inc., Eutechnics Division) (www.alphatechnics.com). Calibration standards are traceable to NIST (US National Institute of Standards and Technology). The kit contains one platinum resistant probe, hand held thermometer, mineral oil, cotton swabs and carrying case.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
FormatKit
For Use With (Equipment)2720 Thermal Cycler, GeneAmp 9700