MicroAmp™ Optical 96-Well Reaction Plate with Barcode
The Applied Biosystems™ MicroAmp™ Optical 96-Well Reaction Plate with Barcode is optimized to provide unmatched temperature accuracy and uniformity forRead more
Have Questions?
Change view
Catalog Number
No. of Wells
Quantity
4306737
96
20 Plates
4326659
96
500 Plates
Catalog number 4306737
Price (USD)/ 20 plates
161.65
Online exclusive
175.00
Save 13.35 (8%)
-
Add to cart
No. of Wells:
96
Quantity:
20 Plates
Price (USD)/ 20 plates
161.65
Online exclusive
175.00
Save 13.35 (8%)
Add to cart
MicroAmp™ Optical 96-Well Reaction Plate with Barcode
Catalog number4306737
Price (USD)/ 20 plates
161.65
Online exclusive
175.00
Save 13.35 (8%)
-
Add to cart
The Applied Biosystems™ MicroAmp™ Optical 96-Well Reaction Plate with Barcode is optimized to provide unmatched temperature accuracy and uniformity for fast, efficient PCR amplification. This plate, constructed from a single rigid piece of polypropylene in a 96-well format, is compatible with Applied Biosystems™ 96-Well Real-Time PCR systems and thermal cyclers.
• Frosted to minimize interfering fluorescence from wells of cycling block • Screened to eliminate auto-fluorescent plates • Unique design provides a barrier to ambient air to help ensure well-to-well temperature uniformity • Each reaction plate includes a unique serialized, eight character number label (barcode) that is user-readable and machine-readable to prevent tracking errors
Oil-Free PCR Amplification MicroAmp™ disposables are optimized to provide unmatched temperature accuracy and uniformity for fast, oil-free PCR amplification. Proprietary MicroAmp™ plates are essential for high-performance PCR and help ensure the greatest temperature and amplification uniformity for your samples.
Specifications
BarcodeIncludes Barcode
Certifications/ComplianceCertified DNA/RNase Free
Packaging20 Plates
Product LineMicroAmp
ColorOptical
Product TypeMicroplate
For Use With (Equipment)Thermal Cycler, Genetic Analyzer, DNA Analyzer
Skirt StyleSemi
MaterialPolypropylene
Volume (Metric) Well0.3 mL
No. of Wells96
Volume (Metric) Working0.2 mL
Quantity20 Plates
Reaction SpeedStandard
Shipping ConditionRoom Temperature
Unit Size20 plates
Contents & Storage
Contains 20 plates.
May be stored at room temperature. With proper storage, plates may be stored indefinitely.