Nalgene™ Utility Funnels
Nalgene™ Utility Funnels
Thermo Scientific™

Nalgene™ Utility Funnels

Replace glass lab funnels with these utility funnels, which are an excellent shatter-proof alternative.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)TopCapacity (Metric)Unit Size
4256-0638150 mm1059 mLCase of 72
4256-023462 mm62 mLCase of 288
4256-0234PK
also known as 4256-0234
62 mm62 mLPack of 12
4256-041498 mm270 mLCase of 144
4256-0414PK
also known as 4256-0414
98 mm270 mLPack of 12
4256-0638PK
also known as 4256-0638
150 mm1059 mLPack of 12
Catalog number 4256-0638
Price (USD)/ Case of 72
1,400.40
Online exclusive
1556.00 
Save 155.60 (10%)
-
Add to cart
Diameter (Metric)Top:
150 mm
Capacity (Metric):
1059 mL
Unit Size:
Case of 72
Price (USD)/ Case of 72
1,400.40
Online exclusive
1556.00 
Save 155.60 (10%)
Add to cart
Nalgene™ Utility Funnels
Catalog number4256-0638
Price (USD)/ Case of 72
1,400.40
Online exclusive
1556.00 
Save 155.60 (10%)
-
Add to cart

Replace glass lab funnels with Thermo Scientific™ Nalgene™ Utility Funnels. These funnels are chemically resistant to a wide variety of lab chemicals and reagents and are shatterproof making them a valuble asset to lab safety programs.

  • 4256-0234 and 4256-0414 are made from shatter-proof, autoclavable polypropylene.
  • 4256-0638 has a large throat opening for faster transfer of materials and is made from shatter-proof high-density polyethylene (Not autoclavable).
Recycling Information:
Nalgene reusable labware products are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually.   Thermo Fisher Scientific encourages recycling Nalgene polypropylene (PP) and High Density Polyethylene (HDPE) labware at end of product life in communities where these materials can be recycled.  Please also recycle the corrugate cartons and any plastic bags (LDPE) included in your product packaging.
Specifications
Diameter (Metric)Top150 mm
Capacity (Metric)1059 mL
Diameter (Metric) Stem48 mm
MaterialHDPE
Height (Metric)175 mm
AutoclavableNot Autoclavable
DescriptionFunnel Utility HDPE 162mm
Length (Metric) Stem66 mm
Quantity12/Pk.; 72/Cs.
Unit SizeCase of 72