Catalog Number | Description |
---|---|
42508918 | RadEye SPRD-KIT, contains 1 ea. RadEye SPRD, Lutetium Test Adapter, Desktop holder + USB connection cable, Software package RadEye.exe + RadEyeSpectra, Holster and spare AAA batteries packaged in a hardened plastic case. |
425082541 | RadEye SPRD-ER Kit |
425082542 | Radeye SPRD-ER Vehicle Kit, contains 1 ea. RadEye SPRD-ER, Lutetium Test Adapter, Vehicle charging kit, Bluetooth adapter, Software package RadEye.exe + RadEyeSpectra, Holster and spare AAA rechargeable batteries |
4250825 | RadEye SPRD-ER Personal Radiation Detector |
4250817 | RadEye SPRD Personal Radiation Detector |
4250827 | RadEye SPRD Personal Radiation Detector |
Respond quickly and decisively to real radiation threats with the new standard in detection and interdiction of illicit radiation. The Thermo Scientific™ RadEye™ SPRD and SPRD-ER Personal Radiation Detectors are handheld radiation monitors that offer a unique combination of detector sensitivity and intelligent alarming to help ensure detection in the most challenging applications while eliminating nuisance alarms. Additionally, the RadEye SPRD and SPRD-ER radiation detectors identify and categorizes specific types of radiation or radioactive material, and can be configured to support your missions or user skills.
By providing significant detector sensitivity for search and find, combined with accurate high dose rate radiation measurement, the RadEye SPRD-ER is a suitable instrument for all users and may be the only radiation detector you need.
View our online training here.