eBioscience™ Phalloidin eFluor™ 570
Product Image
Invitrogen™
eBioscience™ Phalloidin eFluor™ 570
Phalloidin is a peptide toxin isolated from the Amanita phalloides mushroom. Phalloidin selectively binds F-actin and functions to stablilize actinRead more
Have Questions?
Catalog number 41-6559-05
Price (USD)/ 300 tests
460.00
-
Add to cart
Price (USD)/ 300 tests
460.00
Add to cart
eBioscience™ Phalloidin eFluor™ 570
Catalog number41-6559-05
Price (USD)/ 300 tests
460.00
-
Add to cart
Phalloidin is a peptide toxin isolated from the Amanita phalloides mushroom. Phalloidin selectively binds F-actin and functions to stablilize actin polymers by inhibiting the dissociation of actin monomers from the filament ends. Because F-actin polymers compose the cytoskeleton of almost all eukaryotic cells, phalloidin conjugates can be useful probes for visualizing the structure of live or fixed cells using microscopy. Phalloidin eFluor™ 570 can be used to label cells of multiple species.
Phalloidin eFluor™ 570 is supplied as a frozen liquid. To prepare the 1000X stock solution, thaw the vial, add 30 μL high-quality anhydrous DMSO, mix and briefly spin down. The resulting solution (phalloidin DMSO stock) should be aliquoted, protected from light and moisture, and stored at -20°C. To prepare a 1X working solution of phalloidin conjugate, add 1 μL of 1000X phalloidin DMSO stock solution to 1 mL of PBS containing 1% BSA.
Reported Applications
  • Immunocytochemistry
  • Immunohistochemical Staining
  • Microscopy
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Excitation Wavelength Range555/570 nm
Dye TypeeFluor 570
No. of Tests300 Tests
Quantity300 tests
Product LineeBioscience™
Label TypeeFluor
Product TypePhalloidin
SubCellular LocalizationCytoskeleton
Unit Size300 tests