Blocker™ Casein
Blocker™ Casein
Thermo Scientific™

Blocker™ Casein

Blocker Casein is a ready-to-use solution of purified casein for blocking steps in western blot, ELISA, IHC, and nucleic acid detection methods.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
37582Blocker Casein in PBS, 100 mL
37528Blocker Casein in PBS, 1 L
37583Blocker Casein in TBS, 100 mL
37528X3Blocker Casein in PBS, 3 x 1 L
37532Blocker Casein in TBS, 1 L
Catalog number 37582
Price (USD)/ 100 mL
49.65
Online exclusive
53.25 
Save 3.60 (7%)
-
Add to cart
Description:
Blocker Casein in PBS, 100 mL
Request bulk or custom format
Price (USD)/ 100 mL
49.65
Online exclusive
53.25 
Save 3.60 (7%)
Add to cart
Blocker™ Casein
Catalog number37582
Price (USD)/ 100 mL
49.65
Online exclusive
53.25 
Save 3.60 (7%)
-
Add to cart
Blocker Casein is a ready-to-use solution of purified casein for blocking steps in western blot, ELISA, IHC, and nucleic acid detection methods. Blocker Casein is 1% (w/v) casein, which corresponds to the optimal concentration for most applications.
Features of Blocker Casein include:
Purified casein—single-protein blocking buffer provides fewer chances of cross-reaction with assay components than serum or milk solutions
Easy to use—1% casein solution is ready to use; can be diluted further as needed
Safe—stable, thimerosal-free formulation

For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptionBlocker Casein in PBS, 100 mL
Product TypeBlocker Casein
Quantity100 mL
FormLiquid
For Use With (Application)Western Blot, ELISA
Product LineBlocker™
Concentration1X
BufferPBS
Blocking AgentSingle purified protein
Unit Size100 mL
Contents & Storage
Store at 4°C.