1-Step™ Ultra TMB-Blotting Solution
1-Step™ Ultra TMB-Blotting Solution
Thermo Scientific™
1-Step™ Ultra TMB-Blotting Solution
Thermo Scientific 1-Step Ultra TMB-Blotting Solution is an enhanced single-component horseradish peroxidase (HRP) substrate for Western blotting.This precipitating, colorimetric WesternRead more
Have Questions?
Catalog number 37574
Price (USD)/ 250 mL
223.65
Online exclusive
234.00 
Save 10.35 (4%)
-
Add to cart
Request bulk or custom format
Price (USD)/ 250 mL
223.65
Online exclusive
234.00 
Save 10.35 (4%)
Add to cart
1-Step™ Ultra TMB-Blotting Solution
Catalog number37574
Price (USD)/ 250 mL
223.65
Online exclusive
234.00 
Save 10.35 (4%)
-
Add to cart
Thermo Scientific 1-Step Ultra TMB-Blotting Solution is an enhanced single-component horseradish peroxidase (HRP) substrate for Western blotting.
This precipitating, colorimetric Western blot substrate for HRP provides high sensitivity, increased signal-to-noise ratio and low background compared to many other chromogenic substrates. The blotting solution contains soluble TMB (3,3',5,5'-tetramethylbenzidine), which reacts very quickly with horseradish peroxidase enzyme to produce an insoluble dark blue precipitate that does not fade or flake. The substrate is compatible with both nitrocellulose and PVDF membranes. The blotting solution is supplied ready to use with no mixing required.
Features of 1-Step Ultra TMB-Blotting Solution:
Fast—protein bands visible in less than one minute
Fade-resistant—protein bands stable after membrane drying
Sensitive—detection limit similar to Pierce ECL Western Blotting Substrate
Chromogenic—no special equipment needed for visualization; produces dark blue bands
Ready to use—no organic solvents are required to dissolve; no dilution necessary for use
Applications:
• Chromogenic Western blot detection
• Nitrocellulose membrane blots
• PVDF membrane blots
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product TypeSubstrate
Detection MethodColorimetric
FormLiquid
Membrane CompatibilityNitrocellulose, PVDF
Product Line1-Step™
Quantity250 mL
SubstrateTMB
Unit Size250 mL
Contents & Storage
Upon receipt store product at 4°C. Product is shipped at ambient temperature.