SuperBlock™ Blocking Buffer
SuperBlock™ Blocking Buffer
Actual product may vary
Thermo Scientific™

SuperBlock™ Blocking Buffer

SuperBlock Blocking Buffer is an optimized solution containing a single purified glycoprotein that provides incredibly fast and effective blocking for ELISA, IHC, and western blot analysis.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
37545SuperBlock (TBS) Blocking Buffer, dry blend, 5 packs
37580SuperBlock (PBS) Blocking Buffer, 100 mL
37515SuperBlock (PBS) Blocking Buffer, 1 L
37518SuperBlock (PBS) Blocking Buffer, 5 L
37581SuperBlock (TBS) Blocking Buffer, 100 mL
37535SuperBlock (TBS) Blocking Buffer, 1 L
37516SuperBlock T20 (PBS) Blocking Buffer, 1 L
37536SuperBlock T20 (TBS) Blocking Buffer, 1 L
Catalog number 37545
Price (USD)/ 5 packs
187.65
Online exclusive
202.00 
Save 14.35 (7%)
-
Add to cart
Description:
SuperBlock (TBS) Blocking Buffer, dry blend, 5 packs
Request bulk or custom format
Price (USD)/ 5 packs
187.65
Online exclusive
202.00 
Save 14.35 (7%)
Add to cart
SuperBlock™ Blocking Buffer
Catalog number37545
Price (USD)/ 5 packs
187.65
Online exclusive
202.00 
Save 14.35 (7%)
-
Add to cart
SuperBlock Blocking Buffer is an optimized solution containing a single purified glycoprotein that provides incredibly fast and effective blocking for ELISA, IHC, and western blot analysis. Blocking micro-plates, membranes, or tissues with a SuperBlock buffer yields a high signal-to-noise ratio in most detection systems.
Features of SuperBlock Blocking Buffer include:
Blocking time—block membranes in 5 to 10 minutes and ELISA plates in 2 minutes
Applications—ELISA, western blotting, IHC; biotin-free for use with streptavidin system
Stability—store buffer at 4°C for one year; store blocked plates dry for up to 12 months

Blocking micro-plates, membranes, or tissues with SuperBlock buffer yields a high signal-to-noise ratio in most detection systems. The protein-based formulation does not contain any immunoglobulins, albumin, or endogenous biotin, making it compatible in many situations where traditional blocking agents fail. The buffer is especially effective at blocking coated polystyrene microplates (96-well plates) and stabilizing them for drying and storage for later use.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptionSuperBlock (TBS) Blocking Buffer, dry blend, 5 packs
Product TypeBlocking Buffer, dry blend
Quantity5 Pk.
FormPowder
For Use With (Application)ELISA, Western Blot
Product LineSuperBlock™
BufferTBS
Blocking AgentSingle purified protein
Unit Size5 packs
Contents & Storage
Upon receipt store at 4°C. Product is shipped at ambient temperature.