Blocker™ Casein
Blocker™ Casein
Thermo Scientific™

Blocker™ Casein

Blocker Casein is a ready-to-use solution of purified casein for blocking steps in western blot, ELISA, IHC, and nucleic acid detection methods.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
37532Blocker Casein in TBS, 1 L
37582Blocker Casein in PBS, 100 mL
37528Blocker Casein in PBS, 1 L
37583Blocker Casein in TBS, 100 mL
37528X3Blocker Casein in PBS, 3 x 1 L
Catalog number 37532
Price (USD)/ 1 L
172.65
Online exclusive
185.00 
Save 12.35 (7%)
-
Add to cart
Description:
Blocker Casein in TBS, 1 L
Request bulk or custom format
Price (USD)/ 1 L
172.65
Online exclusive
185.00 
Save 12.35 (7%)
Add to cart
Blocker™ Casein
Catalog number37532
Price (USD)/ 1 L
172.65
Online exclusive
185.00 
Save 12.35 (7%)
-
Add to cart
Blocker Casein is a ready-to-use solution of purified casein for blocking steps in western blot, ELISA, IHC, and nucleic acid detection methods. Blocker Casein is 1% (w/v) casein, which corresponds to the optimal concentration for most applications.
Features of Blocker Casein include:
Purified casein—single-protein blocking buffer provides fewer chances of cross-reaction with assay components than serum or milk solutions
Easy to use—1% casein solution is ready to use; can be diluted further as needed
Safe—stable, thimerosal-free formulation

For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptionBlocker Casein in TBS, 1 L
Product TypeBlocker Casein
Quantity1 L
FormLiquid
For Use With (Application)Western Blot, ELISA
Product LineBlocker™
Concentration1X
BufferTBS
Blocking AgentSingle purified protein
Unit Size1 L
Contents & Storage
Store at 4°C.