SuperBlock™ Blocking Buffer
SuperBlock™ Blocking Buffer
Thermo Scientific™

SuperBlock™ Blocking Buffer

SuperBlock Blocking Buffer is an optimized solution containing a single purified glycoprotein that provides incredibly fast and effective blocking for ELISA, IHC, and western blot analysis.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
37516SuperBlock T20 (PBS) Blocking Buffer, 1 L
37580SuperBlock (PBS) Blocking Buffer, 100 mL
37515SuperBlock (PBS) Blocking Buffer, 1 L
37518SuperBlock (PBS) Blocking Buffer, 5 L
37581SuperBlock (TBS) Blocking Buffer, 100 mL
37535SuperBlock (TBS) Blocking Buffer, 1 L
37545SuperBlock (TBS) Blocking Buffer, dry blend, 5 packs
37536SuperBlock T20 (TBS) Blocking Buffer, 1 L
Catalog number 37516
Price (USD)/ 1 L
250.65
Online exclusive
270.00 
Save 19.35 (7%)
-
Add to cart
Description:
SuperBlock T20 (PBS) Blocking Buffer, 1 L
Request bulk or custom format
Price (USD)/ 1 L
250.65
Online exclusive
270.00 
Save 19.35 (7%)
Add to cart
SuperBlock™ Blocking Buffer
Catalog number37516
Price (USD)/ 1 L
250.65
Online exclusive
270.00 
Save 19.35 (7%)
-
Add to cart
SuperBlock Blocking Buffer is an optimized solution containing a single purified glycoprotein that provides incredibly fast and effective blocking for ELISA, IHC, and western blot analysis. Blocking micro-plates, membranes, or tissues with a SuperBlock buffer yields a high signal-to-noise ratio in most detection systems.
Features of SuperBlock Blocking Buffer include:
Blocking time—block membranes in 5 to 10 minutes and ELISA plates in 2 minutes
Applications—ELISA, western blotting, IHC; biotin-free for use with streptavidin system
Stability—store buffer at 4°C for one year; store blocked plates dry for up to 12 months

Blocking micro-plates, membranes, or tissues with SuperBlock buffer yields a high signal-to-noise ratio in most detection systems. The protein-based formulation does not contain any immunoglobulins, albumin, or endogenous biotin, making it compatible in many situations where traditional blocking agents fail. The buffer is especially effective at blocking coated polystyrene microplates (96-well plates) and stabilizing them for drying and storage for later use.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptionSuperBlock T20 (PBS) Blocking Buffer, 1 L
Product TypeBlocking Buffer
Quantity1 L
FormLiquid
For Use With (Application)ELISA, Western Blot
Product LineSuperBlock™
Concentration1X
BufferPBST
Blocking AgentSingle purified protein
Unit Size1 L
Contents & Storage
Upon receipt store at 4°C. Product is shipped at ambient temperature.