Matrix™ 500μL ScrewTop Tubes
Matrix™ 500μL ScrewTop Tubes
Thermo Scientific™
Matrix™ 500μL ScrewTop Tubes
Store samples securely at low temperatures, including vapor phase Liquid Nitrogen with these ScrewTop tubes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberClosure ColorDescriptionPackagingFit for Purpose
3743REDRedV-Bottom ScrewTop TubesBulk, 480 TubesSterile & PCR Ready
3743ColorlessV-Bottom ScrewTop TubesBulk, 480 TubesSterile & PCR Ready
3743YELYellowV-Bottom ScrewTop TubesBulk, 480 TubesSterile & PCR Ready
3743BLUBlueV-Bottom ScrewTop TubesBulk, 480 TubesSterile & PCR Ready
3743GREGreenV-Bottom ScrewTop TubesBulk, 480 TubesSterile & PCR Ready
3743WHIWhiteV-Bottom ScrewTop TubesBulk, 480 TubesSterile & PCR Ready
3743PURPurpleV-Bottom ScrewTop TubesBulk, 480 TubesSterile & PCR Ready
3744ColorlessV-Bottom ScrewTop Tubes5 Latch Racks of 96 TubesSterile & PCR Ready
3744REDRedV-Bottom ScrewTop Tubes5 Latch Racks of 96 TubesSterile & PCR Ready
3744YELYellowV-Bottom ScrewTop Tubes5 Latch Racks of 96 TubesSterile & PCR Ready
3744BLUBlueV-Bottom ScrewTop Tubes5 Latch Racks of 96 TubesSterile & PCR Ready
3744GREGreenV-Bottom ScrewTop Tubes5 Latch Racks of 96 TubesSterile & PCR Ready
3744WHIWhiteV-Bottom ScrewTop Tubes5 Latch Racks of 96 TubesSterile & PCR Ready
3744PURPurpleV-Bottom ScrewTop Tubes5 Latch Racks of 96 TubesSterile & PCR Ready
3745n.a.V-Bottom ScrewTop Tubes, No Caps5 Latch Racks of 96 TubesSterile & PCR Ready
3744AMBn.a.2D tube with no cap5 Latch Racks of 96 TubesSterile & PCR Ready
3743AMBRed2D tube with red cap5 Latch Racks of 96 TubesSterile & PCR Ready
3744-WPColorlessV-Bottom ScrewTop Tubes with white patch5 Latch Racks of 96 TubesSterile & PCR Ready
Catalog number 3743RED
Price (USD)/ Case of 480
397.80
Online exclusive
442.00 
Save 44.20 (10%)
-
Add to cart
Closure Color:
Red
Description:
V-Bottom ScrewTop Tubes
Packaging:
Bulk, 480 Tubes
Fit for Purpose:
Sterile & PCR Ready
Price (USD)/ Case of 480
397.80
Online exclusive
442.00 
Save 44.20 (10%)
Add to cart
Matrix™ 500μL ScrewTop Tubes
Catalog number3743RED
Price (USD)/ Case of 480
397.80
Online exclusive
442.00 
Save 44.20 (10%)
-
Add to cart
Store samples securely at low temperatures, including vapor phase Liquid Nitrogen with Thermo Scientific™ Matrix™ 500μL ScrewTop tubes. These tubes are molded from medical grade resins and feature a permanent, high contrast 2D barcode. Used in combination with Thermo Scientific™ VisionMate™ 2D barcode readers, manual and automated capping equipment, and labeling options, the 2D ScrewTop Tubes meet a range of manual and automated long term storage needs.
Dependable Sample Protection
  • Tubes are constructed of virgin class VI medical grade polypropylene
  • ScrewTop caps feature a soft silicone gasket that conforms to mating surfaces to ensure sample preservation
  • Racks are constructed of sturdy acrylonitrile butadiene styrene (ABS) to protect tubes in case of accidental dropping
  • ScrewTop Tubes are compatible with storage down to -176°C
Secure Tracking
  • Permanent, unique 2D barcode is laser-etched on the bottom of each tube
  • 2D barcode allows you to scan and decode tubes at once without removing them from the rack, and allows data to be associated with each tube
  • Compatible with our range of VisionMate readers
Superior Storage Format
  • ANSI-format latch racks maximize storage space and are compatible with multichannel manual pipettes or automated liquid handling equipment
  • Several capping / decapping options available to manipulate a single tube, 8 tubes, or 96 tubes at a time
Stringent Quality Control
  • Every 2D barcode is scanned for readability
  • Every 2D barcode is checked against the database of all previous Thermo Scientific 2D barcodes to ensure there are zero duplicates
  • Each tube is leak tested to ensure tube integrity and sample security
  • All tubes are supplied free from DNA, RNase, DNase, PCR Inhibitors, endotoxins and cytotoxins
WARNING: Cancer – www.P65Warnings.ca.gov
Specifications
Product TypeScrew Top Tube
DescriptionV-Bottom ScrewTop Tubes
Closure ColorRed
AutoclavableNot Autoclavable
Capacity (Metric)500 μL
Bottom ShapeV-bottom
MaterialPolypropylene (Medical Grade)
PackagingBulk, 480 Tubes
Well ShapeV-bottom
BarcodeNon-proprietary, 12x12 Data-matrix with ECC200 Built-in Error Correction
Product LineMatrix™
SterilitySterile
Closure MaterialVirgin Class VI Medical Grade Polypropylene with Virgin Class VI Medical Grade Silicone O-ring
Capacity (English)16907 fl oz.
Dnase FreeYes
Fit for PurposeSterile & PCR Ready
Unit SizeCase of 480