Matrix™ 2D Barcoded Open-Top Storage Tubes
Matrix™ 2D Barcoded Open-Top Storage Tubes
Thermo Scientific™

Matrix™ 2D Barcoded Open-Top Storage Tubes

Store and track sample material in these 2D barcoded clear polypropylene open-top storage tubes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)SterilityWell ShapeFit for Purpose
37101.4 mLNon-sterileFlatPCR Ready
3719-111.4 mLNon-sterileFlatPCR Ready
3725TS
also known as 3725
0.5 mLSterileV-bottomSterile & PCR Ready
3715-111.4 mLNon-sterileFlatPCR Ready
3730-110.75 mLNon-sterileV-bottomPCR Ready
3731-110.75 mLNon-sterileV-bottomPCR Ready
3712-111.4 mLSterileFlatSterile & PCR Ready
37111.4 mLNon-sterileFlatPCR Ready
37121.4 mLSterileFlatSterile & PCR Ready
37300.75 mLNon-sterileV-bottomSterile & PCR Ready
37320.75 mLSterileV-bottomSterile & PCR Ready
37340.5 mLNon-sterileV-bottomPCR Ready
3735500 μLSterileV-bottomSterile & PCR Ready
3750500 μLNon-sterileV-bottomPCR Ready
37901.4 mLNon-sterileV-bottomPCR Ready
37911.4 mLNon-sterileV-bottomPCR Ready
37921.4 mLSterileV-bottomSterile & PCR Ready
Catalog number 3710
Price (USD)/ Case of 1000
604.35
Online exclusive
711.00 
Save 106.65 (15%)
-
Add to cart
Capacity (Metric):
1.4 mL
Sterility:
Non-sterile
Well Shape:
Flat
Fit for Purpose:
PCR Ready
Price (USD)/ Case of 1000
604.35
Online exclusive
711.00 
Save 106.65 (15%)
Add to cart
Matrix™ 2D Barcoded Open-Top Storage Tubes
Catalog number3710
Price (USD)/ Case of 1000
604.35
Online exclusive
711.00 
Save 106.65 (15%)
-
Add to cart

Store and track samples in Thermo Scientific™ Matrix™ 2D Barcoded Clear Polypropylene Open-Top Storage Tubes. Laser etching produces a permanent, high-contrast, 2D barcode offering maximum reliability and traceability of compound, biological and genomic samples. Offered in three sizes, 0.5mL, 0.75mL, and 1.4mL, these tubes can be sealed with Thermo Scientific SepraSeal septum caps and stored down to -80°C. Item number 3792 is ACT Labeled by My Green Lab to enable data-based sustainable product purchasing decisions.

Secure Tracking:

  • A unique 2D barcode is laser-etched onto the base of every polypropylene open-top tube to securely identify and track samples.
  • 2D barcodes are resistant to 100% DMSO, ideal for compound processing and storage.
  • Non-proprietary 2D codes can be scanned in less than a second in VisionMate High Speed for fast decoding and sample tracking.

Robust Storage Rack Design:

  • Matrix 2D barcoded open-top storage tubes come racked in 96-format in a proprietary specially designed, stackable, ANSI footprint.
  • Latch racks save precious space in your freezer, refrigerator, compound store or on the bench top.
  • The latch rack design enables manual multichannel pipette access to 2D tubes and eliminates contamination risk with lid design that does not contact the benchtop.
  • The latch rack lid can be lifted robotically for 2D tube access with automated liquid handling systems and high throughput applications.

Stringent Quality Control:

  • Every 2D barcoded storage tube is scanned to guarantee readability.
  • Each code is checked against a full database of all previously assigned 2D codes to ensure zero duplicates across the entire line of Matrix 2D barcoded storage tubes.
  • Every storage tube is leak tested to ensure the integrity and security of samples.
  • Matrix storage tubes are manufactured from medical grade virgin polypropylene
  • Tubes are validated to be free of DNA, DNAse, RNAse, PCR Inhibitors, endotoxins and cytotoxins

Storage Tracking Solutions for Any Laboratory:

  • Tubes are available with a range of sealing options including solid and pre-slit Duraseals and Sepraseals to meet your individual storage requirements.
  • Custom 2D coding options are available to meet your laboratory's individual database or tracking requirements.
  • 2D storage tubes are compatible with Thermo Scientific freezer racks that allow optimal utilization of storage space.
  • Item 3710 participates in My Green Lab's ACT Label program for Accountability, Consistency and Transparency (ACT) around evaluating product sustainability

Recommended for:

Individual sample retrieval, instant sample identification, secure identification of archived samples, secure shipping of samples, storage of samples and tracking of samples.

Recycling Information:

Matrix tubes, tube racks and covers are made from polypropylene copolymer and are recyclable in communities where polypropylene is recycled. Please recycle only clean, dry components that have been decontaminated of any hazardous materials or residues. The orange nylon rack latches must be removed from the tube rack base and discarded before recycling the rack base. Tip: Orange latches can be removed with relative ease with the aid of pliers.

Matrix tubes are delivered in high graphic paper cartons. Please recycle the paper cartons and product inserts.

Specifications
Capacity (Metric)1.4 mL
MaterialPolypropylene
PackagingBulk, 1000 Tubes
Well ShapeFlat
Barcode2D barcoded w/human readable code
Product LineMatrix™
SterilityNon-sterile
Capacity (English)0.0473 fl oz.
Fit for PurposePCR Ready
Unit SizeCase of 1000