Nalgene™ Opaque Amber HDPE Diagnostic Bottles without Closure: Bulk Pack
Nalgene™ Opaque Amber HDPE Diagnostic Bottles without Closure: Bulk Pack
Thermo Scientific™

Nalgene™ Opaque Amber HDPE Diagnostic Bottles without Closure: Bulk Pack

Safely store and ship light-sensitive, small-volume, diagnostic samples and valuable reagents with these opaque, amber HDPE bottles.
Have Questions?
Change viewbuttonViewtableView
Capacity (Metric):
4 mL
8 mL
15 mL
Neck Finish:
View all
13-415 mm
Catalog number 362004-9125
Price (USD)/ Case of 2000
825.35
Online exclusive
971.00 
Save 145.65 (15%)
-
Add to cart
Capacity (Metric):
4 mL
Neck Finish:
13-415 mm
Price (USD)/ Case of 2000
825.35
Online exclusive
971.00 
Save 145.65 (15%)
Add to cart
Nalgene™ Opaque Amber HDPE Diagnostic Bottles without Closure: Bulk Pack
Catalog number362004-9125
Price (USD)/ Case of 2000
825.35
Online exclusive
971.00 
Save 145.65 (15%)
-
Add to cart

Safely store and ship light-sensitive, diagnostic samples and valuable reagents in small volumes with Thermo Scientific™ Nalgene™ Opaque Amber HDPE Diagnostic Bottles, used in test kits and many small-volume applications. These opaque, amber, high-density polyethylene bottles are for use with light-sensitive reagents and are packed without closures so they can be matched with various colored closures (Cat. No. 362150 series).

Features:
  • Nonsterile
  • Durable, uniform walls resistant to splitting or puncturing
  • Leakproof† when used with Nalgene closures (must be ordered separately)

Ordering Information:

Bottles are bulk packed without closures. Packed 2,000 per case. Order colored closures separately (Cat. No. 362150).

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

A sample product certificate providing Cytotoxicity, USP Class VI, FDA Compliance—21 CFR and other regulatory compliance information is available or can be requested from our Certificates of Compliance Database

Specifications
Capacity0.125 oz., 4 mL
Capacity (Metric)4 mL
Capacity (English)0.125 oz.
ColorOpaque Amber
Description3.4mL, 20-415 neck finish, bulk pack
Diameter (Metric) Inner Neck8 mm
MaterialHDPE
No. per Case2000
No. per Pack2000
SterilityNon-sterile
Neck Finish13-415 mm
Certifications/ComplianceManufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Classs VI. Product meets the requirements for light-resistant containers per USP latest edition.
Closure IncludedNo
GraduatedNo
MouthNarrow
ShapeRound
Height (Metric)41 mm
Product TypeSample Container
Width (English)0.62 in.
Height (English)1.61 in.
Closure MaterialPolypropylene
Closure Size13-415 mm
Diameter (Metric)16 mm
Unit SizeCase of 2000