Pierce™ Fast Western Kit, SuperSignal™ West Pico, Rabbit
Thermo Scientific™
Pierce™ Fast Western Kit, SuperSignal™ West Pico, Rabbit
Thermo Scientific Pierce Fast Western Blot Kit, SuperSignal West Pico Substrate contains optimized reagents and protocol for picogram-level detection andRead more
Have Questions?
Change view
Catalog Number
Description
35066
Pierce Fast Western Blot Kit, SuperSignal West Pico Substrate, Rabbit Trial Size
35061
Pierce Fast Western Blot Kit, SuperSignal West Pico Substrate, Rabbit
Catalog number 35066
Price (USD)/ 200 mL
144.65
Online exclusive
155.00
Save 10.35 (7%)
-
Add to cart
Description:
Pierce Fast Western Blot Kit, SuperSignal West Pico Substrate, Rabbit Trial Size
Request bulk or custom format
Price (USD)/ 200 mL
144.65
Online exclusive
155.00
Save 10.35 (7%)
Add to cart
Pierce™ Fast Western Kit, SuperSignal™ West Pico, Rabbit
Catalog number35066
Price (USD)/ 200 mL
144.65
Online exclusive
155.00
Save 10.35 (7%)
-
Add to cart
Thermo Scientific Pierce Fast Western Blot Kit, SuperSignal West Pico Substrate contains optimized reagents and protocol for picogram-level detection and and Western blot development less than 1 hour after gel transfer.
Features of this kit:
• Fast—picogram levels of detection in about 1 hour • Convenient—no expensive hardware or vacuum required; no clogging issues • Simple—optimized protocol makes Western blot analysis easier than ever • Economical—cost as little as $14 per blot; no expensive consumables or extra equipment • Excellent stability—kit is stable for 1 year stored at 4°C • Easy—complete kits contain all components needed to block, probe and develop a blot with your rabbit primary antibody
These Fast Western Blot Kits require minimal hands-on time and yield results comparable to classic Western blotting with SuperSignal West Pico Chemiluminescent Substrate.The kits contain all the reagents necessary to complete a Western blot being probed with either a mouse or rabbit primary antibody. Like other Western blotting methods, the included SuperSignal West Pico Substrate produces a signal that is detectable using photographic or other imaging methods. The developed blots can be repeatedly exposed and stripped of the primary antibody and reprobed.
The Pierce Fast Western Blot Kits are reagent-based system that provide optimized reagents for blocking, antibody dilution and detection of Western blots. The Fast Western Blot Kit protocol is quick and efficient method for developing Western blots without the need to purchase expensive instrumentation and consumables required by other popular rapid Western blotting systems.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptionPierce Fast Western Blot Kit, SuperSignal West Pico Substrate, Rabbit Trial Size
Product TypeWestern Blot Kit
Detection MethodChemiluminescence
FormLiquid
For Use With (Application)Western Blot
Membrane CompatibilityNitrocellulose, PVDF
Product LinePierce™, SuperSignal™
Quantity200 mL
ConjugateHRP
SubstrateHorseradish Peroxidase
ReactivityRabbit
SensitivityPicogram-level
Sufficient For5 Mini Blots
Unit Size200 mL
Contents & Storage
Sufficient For: 5 mini blots probed with rabbit primary antibodies • Fast Western Antibody Diluent, 100 mL • Fast Western 10X Wash Buffer, 50 mL • Fast Western Rabbit Optimized HRP Reagent, Pico, 5 mL • SuperSignal West Pico Luminol/Enhancer Solution, 25 mL • SuperSignal West Stable Peroxide Solution, 25 mL