Pierce™ Fast Western Kit, SuperSignal™ West Pico, Mouse
Pierce™ Fast Western Kit, SuperSignal™ West Pico, Mouse
Actual product may vary
Thermo Scientific™

Pierce™ Fast Western Kit, SuperSignal™ West Pico, Mouse

Thermo Scientific Pierce Fast Western Blot Kit, SuperSignal West Pico Substrate contains optimized reagents and protocol for picogram-level detection andRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
35065Pierce Fast Western Blot Kit, SuperSignal West Pico Substrate, Mouse Trial Size
35060Pierce Fast Western Blot Kit, SuperSignal West Pico Substrate, Mouse
Catalog number 35065
Price (USD)/ 200 mL
139.65
Online exclusive
152.00 
Save 12.35 (8%)
-
Add to cart
Description:
Pierce Fast Western Blot Kit, SuperSignal West Pico Substrate, Mouse Trial Size
Request bulk or custom format
Price (USD)/ 200 mL
139.65
Online exclusive
152.00 
Save 12.35 (8%)
Add to cart
Pierce™ Fast Western Kit, SuperSignal™ West Pico, Mouse
Catalog number35065
Price (USD)/ 200 mL
139.65
Online exclusive
152.00 
Save 12.35 (8%)
-
Add to cart
Thermo Scientific Pierce Fast Western Blot Kit, SuperSignal West Pico Substrate contains optimized reagents and protocol for picogram-level detection and and Western blot development less than 1 hour after gel transfer.

Features of this kit:

Fast—picogram levels of detection in about 1 hour
Convenient—no expensive hardware or vacuum required; no clogging issues
Simple—optimized protocol makes Western blot analysis easier than ever
Economical—cost as little as $14 per blot; no expensive consumables or extra equipment
Excellent stability—kit is stable for 1 year stored at 4°C
Easy—complete kits contain all components needed to block, probe and develop a blot with your mouse primary antibody

These Fast Western Blot Kits require minimal hands-on time and yield results comparable to classic Western blotting with SuperSignal West Pico Chemiluminescent Substrate.The kits contain all the reagents necessary to complete a Western blot being probed with either a mouse or rabbit primary antibody. Like other Western blotting methods, the included SuperSignal West Pico Substrate produces a signal that is detectable using photographic or other imaging methods. The developed blots can be repeatedly exposed and stripped of the primary antibody and reprobed.

The Pierce Fast Western Blot Kits are reagent-based system that provide optimized reagents for blocking, antibody dilution and detection of Western blots. The Fast Western Blot Kit protocol is quick and efficient method for developing Western blots without the need to purchase expensive instrumentation and consumables required by other popular rapid Western blotting systems.

Related Products
Pierce™ Fast Western Kit, SuperSignal™ West Pico, Rabbit
For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptionPierce Fast Western Blot Kit, SuperSignal West Pico Substrate, Mouse Trial Size
Product TypeWestern Blot Kit
Detection MethodChemiluminescence
FormLiquid
For Use With (Application)Western Blot
Membrane CompatibilityNitrocellulose, PVDF
Product LinePierce™, SuperSignal™
Quantity200 mL
ConjugateHRP
SubstrateHorseradish Peroxidase
ReactivityMouse
SensitivityPicogram-level
Sufficient For5 Mini Blots
Unit Size200 mL
Contents & Storage
Sufficient For: 5 mini blots probed with mouse primary antibodies
• Fast Western Antibody Diluent, 100 mL
• Fast Western 10X Wash Buffer, 50 mL
• Fast Western Mouse Optimized HRP Reagent, Pico, 5 mL
• SuperSignal West Pico Luminol/Enhancer Solution, 25 mL
• SuperSignal West Stable Peroxide Solution, 25 mL

Store kit at 4°C.