Screw Cap Micro Tubes
Screw Cap Micro Tubes
Thermo Scientific™
Screw Cap Micro Tubes
Choose these tubes, which available in conical and free-standing designs when seeking secure, short-term sample storage in low temperatures.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorDescriptionSterilityFit for PurposeUnit Size
3490Natural2.0ml Screw Cap Tube,Knurl,Skirted,Natural, Non-Sterile tube only no capNon-sterilePCR ReadyCase of 5000
3422NKNatural0.5ml Screw Cap Tube, NonKnurl,Skirted,Natural, Non-Sterile tube only no capNon-sterilePCR ReadyCase of 5000
3472NKNatural0.5ml Screw Cap Tube, NonKnurl,Skirted,Natural, Non-Sterile tube w/ unattached capNon-sterilePCR ReadyCase of 5000
3465NKNatural0.5ml Screw Cap Tube, NonKnurl,Skirted,Natural, E-Beam Sterile tube w/ attached capSterileSterile & PCR ReadyCase of 5000
3422NKSNatural0.5ml Screw Cap Tube, NonKnurl,Skirted,Natural, E-Beam Sterile, tube only no capSterileSterile & PCR ReadyCase of 5000
3462NKNatural0.5ml Screw Cap Tube, NonKnurl, NonSkirted,Natural, Non-Sterile tube only no capNon-sterilePCR ReadyCase of 5000
3460NKNatural0.5ml Screw Cap Tube, NonKnurl, NonSkirted,Natural, Non-Sterile tube w/ unattached capNon-sterilePCR ReadyCase of 5000
3431NKNatural0.5ml Screw Cap Tube, NonKnurl, NonSkirted,Natural, E-Beam Sterile tube w/ attached capSterileSterile & PCR ReadyCase of 5000
3462NKSNatural0.5ml Screw Cap Tube, NonKnurl, NonSkirted,Natural, E-Beam Sterile, tube only no capSterileSterile & PCR ReadyCase of 5000
3422-11Natural0.5ml Screw Cap Tube,Knurl,Skirted,Natural, Non-Sterile tube only no capNon-sterilePCR ReadyCase of 5000
3422-11PK
also known as 3422-11
Natural0.5ml Screw Cap Tube,Knurl,Skirted,Natural, Non-Sterile tube only no capNon-sterilePCR ReadyPack of 500
3472Natural0.5ml Screw Cap Tube,Knurl,Skirted,Natural, Non-Sterile tube w/ unattached capNon-sterilePCR ReadyCase of 5000
3465Natural0.5ml Screw Cap Tube,Knurl,Skirted,Natural, E-Beam Sterile tube w/ attached capSterileSterile & PCR ReadyCase of 5000
3465PKNatural0.5ml Screw Cap Tube,Knurl,Skirted,Natural, E-Beam Sterile tube w/ attached capSterileSterile & PCR ReadyPack of 500
3422SNatural0.5ml Screw Cap Tube,Knurl,Skirted,Natural, E-Beam Sterile, tube only no capSterileSterile & PCR ReadyCase of 5000
3466NKNatural1.5ml Screw Cap Tube, NonKnurl, NonSkirted,Natural, Non-Sterile tube only no capNon-sterilePCR ReadyCase of 5000
3464NKNatural1.5ml Screw Cap Tube, NonKnurl, NonSkirted,Natural, Non-Sterile tube w/ unattached capNon-sterilePCR ReadyCase of 5000
3461NKNatural1.5ml Screw Cap Tube, NonKnurl, NonSkirted,Natural, E-Beam Sterile tube w/ attached capSterileSterile & PCR ReadyCase of 5000
3466NKSNatural1.5ml Screw Cap Tube, NonKnurl, NonSkirted,Natural, E-Beam Sterile, tube only no capSterileSterile & PCR ReadyCase of 5000
3478Natural1.5ml Screw Cap Tube,Knurl,Skirted,Natural, Non-Sterile tube only no capNon-sterilePCR ReadyCase of 5000
3478PK
also known as 3478
Natural1.5ml Screw Cap Tube,Knurl,Skirted,Natural, Non-Sterile tube only no capNon-sterilePCR ReadyPack of 500
3474Natural1.5ml Screw Cap Tube,Knurl,Skirted,Natural, Non-Sterile tube w/ unattached capNon-sterilePCR ReadyCase of 5000
3474PK
also known as 3474
Natural1.5ml Screw Cap Tube,Knurl,Skirted,Natural, Non-Sterile tube w/ unattached capNon-sterilePCR ReadyPack of 500
3467-11Natural1.5ml Screw Cap Tube,Knurl,Skirted,Natural, E-Beam Sterile tube w/ attached capSterileSterile & PCR ReadyCase of 5000
3478SNatural1.5ml Screw Cap Tube,Knurl,Skirted,Natural, E-Beam Sterile, tube only no capSterileSterile & PCR ReadyCase of 5000
3466Natural1.5ml Screw Cap Tube,Knurl, NonSkirted,Natural, Non-Sterile tube only no capNon-sterilePCR ReadyCase of 5000
3466PK
also known as 3466
Natural1.5ml Screw Cap Tube,Knurl, NonSkirted,Natural, Non-Sterile tube only no capNon-sterilePCR ReadyPack of 500
3464Natural1.5ml Screw Cap Tube,Knurl, NonSkirted,Natural, Non-Sterile tube w/ unattached capNon-sterilePCR ReadyCase of 5000
3464PK
also known as 3464
Natural1.5ml Screw Cap Tube,Knurl, NonSkirted,Natural, Non-Sterile tube w/ unattached capNon-sterilePCR ReadyPack of 500
3461Natural1.5ml Screw Cap Tube,Knurl, NonSkirted,Natural, E-Beam Sterile tube w/ attached capSterileSterile & PCR ReadyCase of 5000
3466SNatural1.5ml Screw Cap Tube,Knurl, NonSkirted,Natural, E-Beam Sterile, tube only no capSterileSterile & PCR ReadyCase of 5000
3490NKNatural2.0ml Screw Cap Tube, NonKnurl,Skirted,Natural, Non-Sterile tube only no capNon-sterilePCR ReadyCase of 5000
3488NKNatural2.0ml Screw Cap Tube, NonKnurl,Skirted,Natural, Non-Sterile tube w/ unattached capNon-sterilePCR ReadyCase of 5000
3469NKNatural2.0ml Screw Cap Tube, NonKnurl,Skirted,Natural, E-Beam Sterile tube w/ attached capSterileSterile & PCR ReadyCase of 5000
3490NKSNatural2.0ml Screw Cap Tube, NonKnurl,Skirted,Natural, E-Beam Sterile, tube only no capSterileSterile & PCR ReadyCase of 5000
3470NKNatural2.0ml Screw Cap Tube, NonKnurl, NonSkirted,Natural, Non-Sterile tube only no capNon-sterilePCR ReadyCase of 5000
3468NKNatural2.0ml Screw Cap Tube, NonKnurl, NonSkirted,Natural, Non-Sterile tube w/ unattached capNon-sterilePCR ReadyCase of 5000
3463NKNatural2.0ml Screw Cap Tube, NonKnurl, NonSkirted,Natural, E-Beam Sterile tube w/ attached capSterileSterile & PCR ReadyCase of 5000
3470NKSNatural2.0ml Screw Cap Tube, NonKnurl, NonSkirted,Natural, E-Beam Sterile, tube only no capSterileSterile & PCR ReadyCase of 5000
3490PK
also known as 3490
Natural2.0ml Screw Cap Tube,Knurl,Skirted,Natural, Non-Sterile tube only no capNon-sterilePCR ReadyPack of 500
3488Natural2.0ml Screw Cap Tube,Knurl,Skirted,Natural, Non-Sterile tube w/ unattached capNon-sterilePCR ReadyCase of 5000
3488PK
also known as 3488
Natural2.0ml Screw Cap Tube,Knurl,Skirted,Natural, Non-Sterile tube w/ unattached capNon-sterilePCR ReadyPack of 500
3469-11Natural2.0ml Screw Cap Tube,Knurl,Skirted,Natural, E-Beam Sterile tube w/ attached capSterileSterile & PCR ReadyCase of 5000
3469-11PK
also known as 3469-11
Natural2.0ml Screw Cap Tube,Knurl,Skirted,Natural, E-Beam Sterile tube w/ attached capSterileSterile & PCR ReadyPack of 500
3490SNatural2.0ml Screw Cap Tube,Knurl,Skirted,Natural, E-Beam Sterile, tube only no capSterileSterile & PCR ReadyCase of 5000
3470Natural2.0ml Screw Cap Tube,Knurl, NonSkirted,Natural, Non-Sterile tube only no capNon-sterilePCR ReadyCase of 5000
3470PKNatural2.0ml Screw Cap Tube,Knurl, NonSkirted,Natural, Non-Sterile tube only no capNon-sterilePCR ReadyPack of 500
3468Natural2.0ml Screw Cap Tube,Knurl, NonSkirted,Natural, Non-Sterile tube w/ unattached capNon-sterilePCR ReadyCase of 5000
3468PK
also known as 3468
Natural2.0ml Screw Cap Tube,Knurl, NonSkirted,Natural, Non-Sterile tube w/ unattached capNon-sterilePCR ReadyPack of 500
3463Natural2.0ml Screw Cap Tube,Knurl, NonSkirted,Natural, E-Beam Sterile tube w/ attached capSterileSterile & PCR ReadyCase of 5000
3463PK
also known as 3463
Natural2.0ml Screw Cap Tube,Knurl, NonSkirted,Natural, E-Beam Sterile tube w/ attached capSterileSterile & PCR ReadyPack of 500
3470SNatural2.0ml Screw Cap Tube,Knurl, NonSkirted,Natural, E-Beam Sterile, tube only no capSterileSterile & PCR ReadyCase of 5000
3467TSPK
also known as 3467
NaturalFree-standing tube with screw cap & o-ringSterileSterile & PCR ReadyPack of 500
Catalog number 3490
Price (USD)/ Case of 5000
333.00
Online exclusive
528.00 
Save 195.00 (37%)
-
Add to cart
Color:
Natural
Description:
2.0ml Screw Cap Tube,Knurl,Skirted,Natural, Non-Sterile tube only no cap
Sterility:
Non-sterile
Fit for Purpose:
PCR Ready
Unit Size:
Case of 5000
Price (USD)/ Case of 5000
333.00
Online exclusive
528.00 
Save 195.00 (37%)
Add to cart
Screw Cap Micro Tubes
Catalog number3490
Price (USD)/ Case of 5000
333.00
Online exclusive
528.00 
Save 195.00 (37%)
-
Add to cart
Choose Thermo Scientific™ Screw Cap Micro Tubes, which available in conical and free-standing designs, when seeking secure, short-term sample storage in low temperatures. Their robust design and material selection are well suited for work with biologics and offer rich features, including ultra-clear, externally threaded tubes and UV protective amber tubes. Options include: with or without a self-standing skirted bottom, with or without tethered straps, non-knurl and knurled tubes for single-hand operation, choice of multicolored caps and volume sizes of 0.5mL, 1.5mL and 2.0mL sizes. Screw Cap Micro Tube Caps offer a durable design incorporating an O-ring that helps prevent leaks under demanding conditions.
Non-Sterile is free of Rnase/Dnase, Human and Mouse DNA, Pyrogen & Endotoxin, ATP, PCR Inhibitor.
Sterile E-Beam is free of Rnase/Dnase, Human and Mouse DNA, Pyrogen, Endotoxin, ATP and PCR Inhibitor.
Packaging is tailored to meet broad requirements. Tubes are packaged with and without caps, caps are attached or unattached, placed in a single bag and double bagged feature for clean room environments is available upon request.
100% virgin polypropylene resin specifically selected for high clarity and biologic applications.
O-ring design, thickness and material selection have been optimized to provide protective features for maximum sealing ability, biologic interaction and temperature capability. The black colored O-ring is made from EPDM.
Automated System Compatible. Whether automate is used for filling, capping or labeling, the product is designed to withstand high torque pressures associated with automation. Product design helps to address common automated issues like: fissure cracks, bowl feeder jam, over/under torqued caps and cross threading.
Volume range of 0.5, 1.5 and 2.0mL.
Tube Color choices are an ultra-high clarity, natural color or amber option which provides ultra violet (UV) protection of 1.22% transmittance between 220 and 500nm.
Standard or Tethered cap, all available in a rainbow of colors. Visit cap product page for additional information.
Knurl tubes feature the ability to lock tube into specialty racks for single-hand operation.
Non knurl tubes ideal for automation and labeling, tubes are plain, removed of any labeling interference.
Skirted or Self Standing tubes can stand upright on their own, no rack required.
Conical tubes requires a rack to support them in the upright position.
Low nucleic acid and protein binding and low enzyme inhibition are features that Resin selection features.
RCF range from 18,000 to 25,000 x g.
Temperature Range -80°C to +95°C. Short-term storage in -80°C, -20°C, dry ice and wet ice. Short-term storage in vapor phase LN2. Suitable for a quick dip flash freeze in LN2.
Water Bath compatible up to +95°C.
† Subject to Thermo Fisher Scientific standard limited warranty. See thermoscientific.com or your sales representative for details.
Screw cap micro tubes and caps are not recommended for autoclave. Sterile options available.
Not intended to submerge and store in liquid phase LN2.
Specifications
Max. RCF18,000 x g
Description2.0ml Screw Cap Tube,Knurl,Skirted,Natural, Non-Sterile tube only no cap
AutoclavableNot Autoclavable
Capacity (Metric)2 mL
ColorNatural
MaterialPolypropylene (Virgin)
Packaging500 x pk, 10 pk x cs, 5000 x cs. Single Bagged. ZipLock bag.
Temperature Range (Metric)Cold Storage: -80° to +95°C. Liquid Nitrogen: Short term Vapor Phase & Quick Flash Freeze acceptable, not recommmended for LN2 submersion storage
Clean ClaimsNon-Sterile Free of: Rnase/Dnase, Human and Mouse DNA, Pyrogen & Endotoxin, ATP, PCR Inhibitor
Binding PropertyLow Nucleic Acid and Protein Binding & Low Enzyme Inhibition
Max. TorqueManual/Hand Torque: Standard: 0.125 lbsf.in to 2.5 lbsf.in; Metric: 1.412 Ncm to 28.24 Ncm. Automated/Robotic Torque Standard: 1.312 lbsf.in to 2.5 lbsf.in; Metric: 14.83 Ncm to 28.24 Ncm
SterilityNon-sterile
GraduatedYes
Fit for PurposePCR Ready
Unit SizeCase of 5000